Search
Search
Invitrogen
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}
FIGURE: 1 / 9
Please note: We are reviewing Western blot images included in the antibody testing data in our catalog, including those provided by third parties. Unless expressly labeled or annotated as “raw-unedited”, Western blot images included in the antibody testing data in our catalog may have been edited, optimized or otherwise adjusted for presentation.
Recommended positive controls: 293T, HeLaS3, mouse kidney, rat kidney.
Predicted reactivity: Mouse (99%), Rat (99%), Zebrafish (87%), Xenopus laevis (92%), Pig (99%), Rhesus Monkey (100%), Bovine (99%).
Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
This gene encodes one of seven subunits of the human Arp2/3 protein complex. The Arp2/3 protein complex has been implicated in the control of actin polymerization in cells and has been conserved through evolution. The exact role of the protein encoded by this gene, the p34 subunit, has yet to be determined. Two alternatively spliced variants have been characterized to date. Additional alternatively spliced variants have been described but their full length nature has not been determined.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: 34 kD subunit of the Arp2/3 protein complex; actin related protein 2/3 complex, subunit 2, 34kDa; arp2/3 complex 34 kDa subunit; ARP2/3 protein complex subunit 34; NB4 apoptosis/differentiation related protein; p34-ARC; testis tissue sperm-binding protein Li 53e; unnamed protein product
Gene Aliases: 2210023N03Rik; 34kDa; ARC34; fk84a07; p34-Arc; p34Arc; PNAS-139; PRO2446; wu:fa22f07; wu:fk84a07; zgc:77769
UniProt ID: (Human) O15144, (Mouse) Q9CVB6
Entrez Gene ID: (Human) 10109, (Rat) 301511, (Mouse) 76709, (Zebrafish) 327068
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn more
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support