Search
Search
Invitrogen
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}
FIGURE: 1 / 10
Please note: We are reviewing Western blot images included in the antibody testing data in our catalog, including those provided by third parties. Unless expressly labeled or annotated as “raw-unedited”, Western blot images included in the antibody testing data in our catalog may have been edited, optimized or otherwise adjusted for presentation.
In ICC/IF, autophagosome formation has been seen in HeLa cells after treatment with 50uM chloroquine.
Suggested positive control: human brain lysate, LC3 overexpression lysate.
MAP1A and MAP1B are microtubule-associated proteins which mediate the physical interactions between microtubules and components of the cytoskeleton. MAP1A and MAP1B each consist of a heavy chain subunit and multiple light chain subunits. The protein encoded by this gene is one of the light chain subunits and can associate with either MAP1A or MAP1B. Two transcript variants encoding different isoforms have been found for this gene.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: autophagy-related protein LC3 A; autophagy-related ubiquitin-like modifier LC3 A; LC3-I; LC3-like protein 1; lc3a; LC3I; MAP1 light chain 3-like protein 1; MAP1A/1B light chain 3 A; MAP1A/MAP1B LC3 A; MAP1A/MAP1B light chain 3 A; Microtubule-asso; microtubule-associated protein 1-light chain 3A; microtubule-associated proteins 1A/1B light chain 3; microtubule-associated proteins 1A/1B light chain 3A; MLP3A; RP11-346K17.1; unnamed protein product
Gene Aliases: 1010001H21Rik; 4922501H04Rik; ATG8E; LC3; LC3-I; LC3-II; LC3A; MAP1ALC3; MAP1BLC3; zgc:77094
UniProt ID: (Human) Q9H492, (Bovine) Q2HJ23, (Rat) Q6XVN8, (Mouse) Q91VR7
Entrez Gene ID: (Human) 84557, (Bovine) 514547, (Rat) 362245, (Mouse) 66734, (Zebrafish) 406466
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn more
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support