Search
Search
Invitrogen
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}
FIGURE: 1 / 2
Please note: We are reviewing Western blot images included in the antibody testing data in our catalog, including those provided by third parties. Unless expressly labeled or annotated as “raw-unedited”, Western blot images included in the antibody testing data in our catalog may have been edited, optimized or otherwise adjusted for presentation.
Sequence homology: Mouse, Rat (77%).
FMN2, a novel formin homology protein of the cappuccino subfamily, is highly conserved between evolutionarily diverse vertebrates. FMN2 bears a high degree of similarity to formin-1 which is the founding member of formin homology proteins. FMN2 plays a crucial role in maintenance of the meiotic spindle. The protein is widely expressed in human fetal brain, adult whole brain, hypothalamus, retina, pancreatic islet and germinal-center B cells. Pathological role of the protein is identified in various tumors like parathyloid tumor, glioblastoma, retinoblastoma and chondrosarcoma. FMN2 mutations are often a cause of unexplained infertility in women. The gene encoding for human FMN2 is localized in the chromosomal region 1q43.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: RP11-90L13.1; unnamed protein product
UniProt ID: (Human) Q9NZ56
Entrez Gene ID: (Human) 56776
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn more
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support