Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          • Primary Antibodies ›
          • HEY2 Antibodies

          Invitrogen

          HEY2 Polyclonal Antibody

          Advanced Verification
          1 Published Figure
          1 Reference
          View all (10) HEY2 antibodies

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}

          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Tech Support
          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Tech Support
          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Tech Support

          Cite HEY2 Polyclonal Antibody

          Additional Information:
          {{banner.description}}
          {{$ctrl.isSkuNotesCollapsed ? 'View more' : 'View less'}}
          • Antibody Testing Data (3)
          • Published Figures (1)
          • Advanced Verification (1)
          HEY2 Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          Group 53 Created with Sketch.
          HEY2 Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          Group 53 Created with Sketch.

          FIGURE: 1 / 5

          {{ $ctrl.currentElement.advancedVerification.fullName }}
          {{ $ctrl.currentElement.advancedVerification.text }}

          HEY2 Antibody (PA5-25807) in IHC (P)

          Immunohistochemistry analysis of HEY2 in formalin fixed and paraffin embedded human brain tissue. Samples were incubated with HEY2 polyclonal antibody (Product # PA5-25807) followed by peroxidase conjugation of the secondary antibody and DAB staining. This data demonstrates the use of this antibody for immunohistochemistry. Clinical relevance has not been evaluated. {{ $ctrl.currentElement.advancedVerification.fullName }} validation info. View more
          Published figure supplied by benchsci-logo
          PMID: {{$ctrl.currentElement.benchSciPubmedId}}
          View Product

          {{$ctrl.videoDesc}}

          {{$ctrl.videoLongDesc}}

          HEY2 Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          HEY2 Antibody in Western Blot (WB)
          HEY2 Antibody in Western Blot (WB)
          HEY2 Antibody in Western Blot (WB)
          HEY2 Antibody
          HEY2 Polyclonal Antibody

          Product Details

          PA5-25807

          Applications
          Tested Dilution
          Publications

          Western Blot (WB)

          1:1,000
          View 1 publication 1 publication

          Immunohistochemistry (Paraffin) (IHC (P))

          1:50-1:100
          -
          Product Specifications

          Species Reactivity

          Human, Mouse

          Host/Isotype

          Rabbit / IgG

          Class

          Polyclonal

          Type

          Antibody

          Immunogen

          KLH conjugated synthetic peptide between 65-91 amino acids from the N-terminal region of human HEY2
          View immunogen

          Conjugate

          Unconjugated Unconjugated Unconjugated

          Form

          Liquid

          Concentration

          0.46 mg/mL

          Purification

          Protein A, Antigen affinity chromatography

          Storage buffer

          PBS, pH 7.4

          Contains

          0.09% sodium azide

          Storage conditions

          Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.

          Shipping conditions

          Ambient (domestic); Wet ice (international)

          RRID

          AB_2543307

          Product Specific Information

          This antibody is predicted to react with bovine, canine, Xenopus and zebrafish based on sequence homology.

          Target Information

          Downstream effector of Notch signaling which may be required for cardiovascular development. Transcriptional repressor which binds preferentially to the canonical E box sequence 5'-CACGTG-3'. Represses transcription by the cardiac transcriptional activators GATA4 and GATA6.

          For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.

          Bioinformatics

          Protein Aliases: bHLHb32; cardiovascular basic helix-loop-helix factor 1; Cardiovascular helix-loop-helix factor 1; CHF1; Class B basic helix-loop-helix protein 32; h hHRT2; hairy and enhancer of split related 2; Hairy and enhancer of split-related protein 2; Hairy-E(spl)-related with YRPW motif 2; Hairy-related transcription factor 2; hairy/enhancer-of-split related with YRPW motif 2; Hairy/enhancer-of-split related with YRPW motif protein 2; hCHF1; HES-related repressor protein 1; HES-related repressor protein 2; HESR-2; hGRL; basic helix-loop-helix transcription factor; hHRT2; HRT-2; HRT2; mHRT2; Protein gridlock homolog; transcription factor; unnamed protein product

          View more View less

          Gene Aliases: BHLHB32; CHF1; GRIDLOCK; GRL; HERP; HERP1; HESR2; HEY2; HRT2

          View more View less

          UniProt ID: (Human) Q9UBP5, (Mouse) Q9QUS4

          View more View less

          Entrez Gene ID: (Human) 23493, (Mouse) 15214

          View more View less

          Function(s)
          RNA polymerase II core promoter proximal region sequence-specific DNA binding RNA polymerase II transcription factor activity, sequence-specific DNA binding core promoter proximal region sequence-specific DNA binding transcriptional repressor activity, RNA polymerase II transcription regulatory region sequence-specific binding DNA binding transcription factor activity, sequence-specific DNA binding protein binding identical protein binding histone deacetylase binding sequence-specific DNA binding protein dimerization activity RNA polymerase II sequence-specific DNA binding transcription factor binding sequence-specific double-stranded DNA binding transcription factor activity, RNA polymerase II core promoter sequence-specific transcription factor activity, protein binding RNA polymerase II activating transcription factor binding transcription factor binding microsatellite binding basic helix-loop-helix transcription factor
          Process(es)
          negative regulation of transcription from RNA polymerase II promoter blood vessel development vasculogenesis muscular septum morphogenesis outflow tract morphogenesis cardiac conduction system development atrioventricular valve development aortic valve morphogenesis pulmonary valve morphogenesis tricuspid valve morphogenesis tricuspid valve formation epithelial to mesenchymal transition involved in endocardial cushion formation endocardial cushion to mesenchymal transition involved in heart valve formation cardiac ventricle morphogenesis cardiac left ventricle morphogenesis cardiac right ventricle morphogenesis ventricular trabecula myocardium morphogenesis cardiac muscle hypertrophy regulation of transcription, DNA-templated regulation of transcription from RNA polymerase II promoter Notch signaling pathway pattern specification process heart development anatomical structure morphogenesis anterior/posterior axis specification anterior/posterior pattern specification positive regulation of heart rate regulation of gene expression negative regulation of gene expression cardiac muscle cell apoptotic process negative regulation of cardiac muscle cell apoptotic process mesenchymal cell development cardiac muscle hypertrophy in response to stress ascending aorta morphogenesis dorsal aorta morphogenesis umbilical cord morphogenesis cell fate commitment regulation of auditory receptor cell differentiation negative regulation of Notch signaling pathway negative regulation of transcription, DNA-templated positive regulation of transcription from RNA polymerase II promoter regulation of neurogenesis smooth muscle cell differentiation negative regulation of smooth muscle cell differentiation ventricular cardiac muscle cell development cardiac muscle cell proliferation positive regulation of cardiac muscle cell proliferation cardiac epithelial to mesenchymal transition heart trabecula formation cardiac septum morphogenesis ventricular septum morphogenesis atrial septum morphogenesis negative regulation of transcription initiation from RNA polymerase II promoter labyrinthine layer blood vessel development blood vessel endothelial cell differentiation artery development arterial endothelial cell differentiation cardiac vascular smooth muscle cell development coronary vasculature morphogenesis pulmonary artery morphogenesis protein-DNA complex assembly negative regulation of biomineral tissue development circulatory system development cochlea development vascular smooth muscle cell development negative regulation of cardiac vascular smooth muscle cell differentiation regulation of vasculogenesis transcription, DNA-templated transcription from RNA polymerase II promoter multicellular organism development Notch signaling involved in heart development negative regulation of transcription regulatory region DNA binding negative regulation of transcription from RNA polymerase II promoter involved in smooth muscle cell differentiation
          It has to be done as per old AB suggested Products section.

          Disclaimer

          Close

          Clicking the images or links will redirect you to a website hosted by BenchSci that provides third-party scientific content. Neither the content nor the BenchSci technology and processes for selection have been evaluated by us; we are providing them as-is and without warranty of any kind, including for use or application of the Thermo Fisher Scientific products presented.

          guarantee_icon

          Performance Guarantee

          If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*

          Learn more
          help_icon

          We're here to help

          Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.

          Contact tech support
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Responsibility Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          México flag icon
          México

          Your items have has been added!


          Host server : magellan-search-green-7d94cb4b65-4lm45:80/100.66.75.98:80.
          git-commit: c9e08c96761173abe34e68f880379696776a4827
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.44.1-2026.02.97.1.0