Search
Search
Invitrogen
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}
FIGURE: 1 / 10
Please note: We are reviewing Western blot images included in the antibody testing data in our catalog, including those provided by third parties. Unless expressly labeled or annotated as “raw-unedited”, Western blot images included in the antibody testing data in our catalog may have been edited, optimized or otherwise adjusted for presentation.
Recommended positive controls: Jurkat, Raji, NCI-H929, Neuro 2A, C8D30, NIH-3T3, Raw264.7, C2C12.
Predicted reactivity: Mouse (96%), Rat (95%), Zebrafish (88%), Xenopus laevis (89%), Rhesus Monkey (99%), Bovine (93%).
Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
The protein encoded by this gene is one of the highly conserved mini-chromosome maintenance proteins (MCM) that are essential for the initiation of eukaryotic genome replication. The hexameric protein complex formed by MCM proteins is a key component of the pre-replication complex (pre_RC) and may be involved in the formation of replication forks and in the recruitment of other DNA replication related proteins. The MCM complex consisting of this protein and MCM2, 6 and 7 proteins possesses DNA helicase activity, and may act as a DNA unwinding enzyme. The phosphorylation of this protein by CDC2 kinase reduces the DNA helicase activity and chromatin binding of the MCM complex. This gene is mapped to a region on the chromosome 8 head-to-head next to the PRKDC/DNA-PK, a DNA-activated protein kinase involved in the repair of DNA double-strand breaks. Alternatively spliced transcript variants encoding the same protein have been reported.
⚠WARNING: This product can expose you to chemicals including mercury, which is known to the State of California to cause birth defects or other reproductive harm. For more information go to www.P65Warnings.ca.gov.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: CDC21 homolog; homolog of S. pombe cell devision cycle 21; MCM4 minichromosome maintenance deficient 4, mitotin; mini chromosome maintenance deficient 4 homolog; minichromosome maintenance deficient 4; minichromosome maintenance deficient 4 homolog; P1-CDC21; unnamed protein product
Gene Aliases: 19G; cb1025; CDC21; CDC54; fc12c09; fj85g09; hCdc21; hm:zeh1616; IMD54; mcdc21; Mcmd4; mKIAA4003; NKCD; NKGCD; P1-CDC21; wu:fc12c09; wu:fj85g09; zeh1616
UniProt ID: (Human) P33991, (Mouse) P49717
Entrez Gene ID: (Human) 4173, (Mouse) 17217, (Zebrafish) 337598
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn more
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support