Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • New Products
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          • Primary Antibodies ›
          • NGF Antibodies

          Invitrogen

          NGF Polyclonal Antibody

          Advanced Verification
          4 Published Figures
          3 References
          View all (60) NGF antibodies

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}

          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Tech Support
          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Tech Support
          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Tech Support

          Cite NGF Polyclonal Antibody

          {{$ctrl.translations['antibody.pdp.skuNotes.additionalInformation'] || 'Additional Information'}}: {{$ctrl.translations['antibody.pdp.skuNotes.additionalInformation']}}:

          {{$ctrl.translations[$ctrl.isSkuNotesCollapsed ? 'antibody.pdp.skuNotes.viewMore' : 'antibody.pdp.skuNotes.viewLess']}}
          • Antibody Testing Data (2)
          • Published Figures (4)
          • Advanced Verification (1)
          NGF Antibody in Western Blot (WB)
          Group 53 Created with Sketch.
          NGF Antibody in Western Blot (WB)
          Group 53 Created with Sketch.

          FIGURE: 1 / 7

          {{ $ctrl.currentElement.advancedVerification.fullName }}
          {{ $ctrl.currentElement.advancedVerification.text }}

          NGF Antibody (PA5-29425) in WB

          Western blot was performed using Anti-NGF Polyclonal Antibody (Product # PA5-29425) and a 30 kDa band corresponding to pro-NGF was observed across in Rat Salivary Gland and not in Rat Brain and Mouse Salivary Gland. Whole cell extracts (30 µg lysate) of Mouse Salivary Gland (Lane 1), Rat Salivary Gland (Lane 2) and Rat Brain (Lane 3) were electrophoresed using NuPAGE™ 12% Bis-Tris Protein Gel (Product # NP0342BOX). Resolved proteins were then transferred onto a Nitrocellulose membrane (Product # IB23002) by iBlot® 2 Dry Blotting System (Pr... View More {{ $ctrl.currentElement.advancedVerification.fullName }} validation info. View more
          Published figure supplied by benchsci-logo
          PMID: {{$ctrl.currentElement.benchSciPubmedId}}
          View Product

          {{$ctrl.videoDesc}}

          {{$ctrl.videoLongDesc}}

          NGF Antibody in Western Blot (WB)
          NGF Antibody in Western Blot (WB)
          NGF Antibody in Western Blot (WB)
          NGF Antibody in Western Blot (WB)
          NGF Antibody in Western Blot (WB)
          NGF Antibody in Western Blot (WB)
          NGF Antibody

          Please note: We are reviewing Western blot images included in the antibody testing data in our catalog, including those provided by third parties. Unless expressly labeled or annotated as “raw-unedited”, Western blot images included in the antibody testing data in our catalog may have been edited, optimized or otherwise adjusted for presentation.

          NGF Polyclonal Antibody

          Product Details

          PA5-29425

          Applications
          Tested Dilution
          Publications

          Western Blot (WB)

          1:500-1:3,000
          View 3 publications 3 publications
          Product Specifications

          Species Reactivity

          Rat, Human, Mouse

          Published species

          Rat

          Host/Isotype

          Rabbit / IgG

          Class

          Polyclonal

          Type

          Antibody

          Immunogen

          Full length human NGF beta Recombinant protein.
          3D Epitope / Immunogen

          Conjugate

          Unconjugated Unconjugated Unconjugated

          Form

          Liquid

          Concentration

          0.91 mg/mL

          Amount

          91 µg

          Purification

          Antigen affinity chromatography

          Storage buffer

          0.1M tris glycine, pH 7, with 20% glycerol

          Contains

          0.01% thimerosal

          Storage conditions

          Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.

          Shipping conditions

          Ambient (domestic); Wet ice (international)

          RRID

          AB_2546901

          Product Specific Information

          Recommended positive controls: NT2D1.

          Predicted reactivity: Mouse (85%), Rat (85%), Dog (90%), Chimpanzee (99%), Bovine (90%).

          Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.

          Target Information

          Nerve growth factor (NGF) influences the survival and differentiation of a variety of neuronal and nonneuronal cells. The actions of this polypeptide result from binding to specific cell surface receptors which are present as both high and low affinity sites. Although both types of receptor bind NGF only the high affinity form is capable of signal transduction. Cross-linking studies with 125I-NGF have shown that the high affinity receptor of rat PC12 cells forms an Mr 158,000 complex with NGF while the low affinity receptor forms an Mr 100,000 complex. Full length cDNA clones for both the human and rat NGF receptor genes have been produced and sequenced, revealing greater than 90% homology between the two species. NGF exhibits nerve growth stimulating activity and is involved in the regulation of growth and the differentiation of sympathetic and certain sensory neurons. Further, NGF stimulates division and differentiation of sympathetic and embryonic sensory neurons. Mutations in the NGF gene have been associated with hereditary sensory and autonomic neuropathy, type 5 (HSAN5), and dysregulation of NGF gene expression is associated with allergic rhinitis.

          ⚠WARNING: This product can expose you to chemicals including mercury, which is known to the State of California to cause birth defects or other reproductive harm. For more information go to www.P65Warnings.ca.gov.

          For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.

          Bioinformatics

          Protein Aliases: B nerve growth factor; b-NGF; beta ngf; BetaNGF; bNGF; MGC161426; MGC161428; Nerve growth factor; nerve growth factor (beta polypeptide); nerve growth factor, beta; nerve growth factor, beta subunit; NGF; NGF b; NGF β; NGFB; NGFbeta; NGFβ; prepro-beta-nerve growth factor; pro-nerve growth factor long; unnamed protein product; β nerve growth factor; β NGF; βNGF

          View more View less

          Gene Aliases: Beta-NGF; HSAN5; NGFB

          View more View less

          UniProt ID: (Human) P01138, (Mouse) P01139, (Rat) P25427

          View more View less

          Entrez Gene ID: (Human) 4803, (Mouse) 18049, (Rat) 310738

          View more View less

          Function(s)
          receptor binding nerve growth factor receptor binding protein binding growth factor activity metalloendopeptidase inhibitor activity cysteine-type endopeptidase activator activity involved in apoptotic process transmembrane receptor protein tyrosine kinase activator activity death receptor agonist activity
          Process(es)
          positive regulation of protein phosphorylation transmembrane receptor protein tyrosine kinase signaling pathway peripheral nervous system development memory circadian rhythm cell proliferation positive regulation of cell proliferation adult locomotory behavior extrinsic apoptotic signaling pathway via death domain receptors positive regulation of gene expression positive regulation of neuron projection development positive regulation of neuron maturation sensory perception of pain nerve development cell proliferation in forebrain neuron differentiation positive regulation of cell growth neuron projection development positive regulation of protein ubiquitination positive regulation of protein autophosphorylation nerve growth factor signaling pathway negative regulation of programmed cell death negative regulation of neuron apoptotic process regulation of neuron differentiation positive regulation of neuron differentiation positive regulation of axon extension positive regulation of transcription from RNA polymerase II promoter positive regulation of Ras protein signal transduction regulation of neurotransmitter secretion neurotrophin TRK receptor signaling pathway positive regulation of collateral sprouting neuron projection morphogenesis modulation of synaptic transmission regulation of release of sequestered calcium ion into cytosol positive regulation of neurotrophin TRK receptor signaling pathway neuron apoptotic process positive regulation of protein kinase B signaling sensory perception of hot stimulus positive regulation of ERK1 and ERK2 cascade extrinsic apoptotic signaling pathway in absence of ligand positive regulation of stem cell proliferation negative regulation of type B pancreatic cell apoptotic process spermatogenesis response to xenobiotic stimulus response to mechanical stimulus response to ozone positive regulation of protein binding response to lipopolysaccharide response to nicotine response to peptide hormone response to glucocorticoid response to electrical stimulus negative regulation of endoplasmic reticulum stress-induced intrinsic apoptotic signaling pathway signal transduction negative regulation of cell proliferation
          It has to be done as per old AB suggested Products section.

          Disclaimer

          Close

          Clicking the images or links will redirect you to a website hosted by BenchSci that provides third-party scientific content. Neither the content nor the BenchSci technology and processes for selection have been evaluated by us; we are providing them as-is and without warranty of any kind, including for use or application of the Thermo Fisher Scientific products presented.

          guarantee_icon

          Performance Guarantee

          If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*

          Learn more
          help_icon

          We're here to help

          Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.

          Contact tech support
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Responsibility Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          México flag icon
          México

          Your items have has been added!


          Host server : magellan-search-blue-6b866bbc8c-mwps6:80/100.66.76.64:80.
          git-commit: 9d76681b284ad04e8ddcd97b79462898b32e8e5b
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.61.1-2026.10.99-1.0