Search
Search
Invitrogen
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}
FIGURE: 1 / 7
Please note: We are reviewing Western blot images included in the antibody testing data in our catalog, including those provided by third parties. Unless expressly labeled or annotated as “raw-unedited”, Western blot images included in the antibody testing data in our catalog may have been edited, optimized or otherwise adjusted for presentation.
Recommended positive controls: Raji, K562, HL-60.
Predicted reactivity: Mouse (99%), Rat (99%), Zebrafish (87%), Rhesus Monkey (99%), Bovine (100%).
Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
Acts as one of several non-catalytic accessory components of the cytoplasmic dynein 1 complex that are thought to be involved in linking dynein to cargos and to adapter proteins that regulate dynein function. Cytoplasmic dynein 1 acts as a motor for the intracellular retrograde motility of vesicles and organelles along microtubules. The intermediate chains mediate the binding of dynein to dynactin via its 150 kDa component (p150-glued) DCNT1. May play a role in mediating the interaction of cytoplasmic dynein with membranous organelles and kinetochores.
⚠WARNING: This product can expose you to chemicals including mercury, which is known to the State of California to cause birth defects or other reproductive harm. For more information go to www.P65Warnings.ca.gov.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: cytoplasmic dynein intermediate chain 1; DH IC-1; DNCIC1; Dync1i1A; Dync1i1B; Dync1i1C; Dync1i1D; dynein cytoplasmic 1 intermediate chain 1A; dynein cytoplasmic 1 intermediate chain 1B; dynein cytoplasmic 1 intermediate chain 1C; dynein cytoplasmic 1 intermediate chain 1D; dynein intermediate chain 1, cytosolic; Dynein intermediate chain 1, cytosolic (DH IC-1) (Cytoplasmic dynein intermediate chain 1); dynein, cytoplasmic, intermediate chain 1; dynein, cytoplasmic, intermediate polypeptide 1; unnamed protein product
Gene Aliases: DH IC-1; DHIC-1; DIC; DNCI1; DNCIC1; IC74; IC74-1A; IC74-1B
UniProt ID: (Human) O14576, (Rat) Q63100, (Mouse) O88485
Entrez Gene ID: (Human) 1780, (Rat) 29564, (Mouse) 13426
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn more
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support