Search
Search
Invitrogen
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}
FIGURE: 1 / 8
Please note: We are reviewing Western blot images included in the antibody testing data in our catalog, including those provided by third parties. Unless expressly labeled or annotated as “raw-unedited”, Western blot images included in the antibody testing data in our catalog may have been edited, optimized or otherwise adjusted for presentation.
Recommended positive controls: A549, mouse liver, mouse kidney.
Predicted reactivity: Mouse (88%), Rat (88%), Chicken (81%), Rhesus Monkey (95%), Chimpanzee (98%), Bovine (90%).
Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
This gene encodes the last enzyme in the tyrosine catabolism pathway. FAH deficiency is associated with Type 1 hereditary tyrosinemia (HT).
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: beta-diketonase; epididymis secretory sperm binding protein; FAA; fumarylacetoacetate hydrolase (fumarylacetoacetase); unnamed protein product
Gene Aliases: swst
UniProt ID: (Human) P16930, (Rat) P25093, (Mouse) P35505
Entrez Gene ID: (Human) 2184, (Rat) 29383, (Mouse) 14085
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn more
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support