Search
Search
Invitrogen
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}
Please note: We are reviewing Western blot images included in the antibody testing data in our catalog, including those provided by third parties. Unless expressly labeled or annotated as “raw-unedited”, Western blot images included in the antibody testing data in our catalog may have been edited, optimized or otherwise adjusted for presentation.
Recommended positive controls: IMR32.
Predicted reactivity: Human (99%), Mouse (84%), Rat (82%), Bovine (89%).
Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
The LRRC23 (Leucine-Rich Repeat Containing 23) gene encodes a protein that is an evolutionarily conserved component of the radial spoke complex in sperm flagella. It is located on human chromosome 12. The LRRC23 protein interacts with radial spoke head proteins such as RSPH3A and RSPH3B, crucial for the structural integrity and proper function of the radial spokes, which are essential for flagellar motility. Mutations or deficiencies in LRRC23 have been linked to male infertility, specifically causing asthenozoospermia, characterized by reduced sperm motility. Studies have identified various mutations in LRRC23 that result in disorganized radial spokes and impaired sperm motility, leading to infertility
⚠WARNING: This product can expose you to chemicals including mercury, which is known to the State of California to cause birth defects or other reproductive harm. For more information go to www.P65Warnings.ca.gov.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: leucine-rich protein B7; unnamed protein product
Gene Aliases: LRPB7; SPGF92
UniProt ID: (Human) Q53EV4
Entrez Gene ID: (Human) 10233
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn more
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support