Search
Search
Invitrogen
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}
Please note: We are reviewing Western blot images included in the antibody testing data in our catalog, including those provided by third parties. Unless expressly labeled or annotated as “raw-unedited”, Western blot images included in the antibody testing data in our catalog may have been edited, optimized or otherwise adjusted for presentation.
Percent identity with other species by BLAST analysis: Human, Gorilla, Gibbon, Marmoset (100%) Hamster, Opossum (86%).
DSCD75 is a 208 amino acid membrane protein with one transmembrane helix. Current testing suggests that the cDNA of DSCD75 may be expressed by osteogenic human mesenchymal stem cells. DSCD75 may be involved in some transporter activities.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: mesenchymal stem cell protein DSCD75; unnamed protein product; UPF0670 protein C8orf55; UPF0670 protein THEM6
Gene Aliases: C8orf55; DSCD75
UniProt ID: (Human) Q8WUY1
Entrez Gene ID: (Human) 51337
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn more
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support