Search
Search
Invitrogen
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}
Please note: We are reviewing Western blot images included in the antibody testing data in our catalog, including those provided by third parties. Unless expressly labeled or annotated as “raw-unedited”, Western blot images included in the antibody testing data in our catalog may have been edited, optimized or otherwise adjusted for presentation.
A suggested positive control is rat brain tissue lysate.
PA5-34545 can be used with blocking peptide PEP-1588.
Defects in the CISD2 (CDGSH iron sulfur domain 2) gene are a cause of the neurodegenerative disorder Wolfram syndrome 2. CISD2 is a zinc finger protein that localizes to the endoplasmic reticulum and mitochondria and binds an iron/sulfur cluster. CISD2 interacts with Bcl-2 and can be displaced by the BH3-only protein BIK and contributes to the regulation of BIK-initiated autophagy. CISD2 deficiency in mice causes mitochondrial breakdown accompanied by autophagic cell death as well as the development of premature aging phenotype.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: endoplasmic reticulum intermembrane small protein; mitoNEET related 1; mitoNEET-related 1 protein; nervous system over-expressed protein 70; nervous system overexpressed protein 70; nutrient-deprivation autophagy factor-1; unnamed protein product; zinc finger, CDGSH-type domain 2
Gene Aliases: 1500009M05Rik; 1500026J14Rik; 1500031D15Rik; B630006A20Rik; ERIS; Miner1; NAF-1; Noxp70; RGD1566242; WFS2; ZCD2
UniProt ID: (Human) Q8N5K1, (Mouse) Q9CQB5
Entrez Gene ID: (Human) 493856, (Mouse) 67006, (Rat) 295457
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn more
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support