Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • New Products
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          • Primary Antibodies ›
          • TIP60 Antibodies

          Invitrogen

          TIP60 Polyclonal Antibody

          View all (20) TIP60 antibodies

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}

          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Tech Support
          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Tech Support
          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Tech Support

          Cite TIP60 Polyclonal Antibody

          {{$ctrl.translations['antibody.pdp.skuNotes.additionalInformation'] || 'Additional Information'}}: {{$ctrl.translations['antibody.pdp.skuNotes.additionalInformation']}}:

          {{$ctrl.translations[$ctrl.isSkuNotesCollapsed ? 'antibody.pdp.skuNotes.viewMore' : 'antibody.pdp.skuNotes.viewLess']}}
          • Antibody Testing Data (4)
          TIP60 Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          Group 53 Created with Sketch.
          TIP60 Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          Group 53 Created with Sketch.

          FIGURE: 1 / 4

          {{ $ctrl.currentElement.advancedVerification.fullName }}
          {{ $ctrl.currentElement.advancedVerification.text }}

          TIP60 Antibody (PA5-34794) in IHC (P)

          TIP60 Polyclonal Antibody detects TIP60 protein at nucleus by immunohistochemical analysis. Sample: Paraffin-embedded mouse duodenum. TIP60 stained by TIP60 Polyclonal Antibody (Product # PA5-34794) diluted at 1:1,000. Antigen Retrieval: Citrate buffer, pH 6.0, 15 min. {{ $ctrl.currentElement.advancedVerification.fullName }} validation info. View more
          Published figure supplied by benchsci-logo
          PMID: {{$ctrl.currentElement.benchSciPubmedId}}
          View Product

          {{$ctrl.videoDesc}}

          {{$ctrl.videoLongDesc}}

          TIP60 Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          TIP60 Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          TIP60 Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          TIP60 Antibody in Western Blot (WB)

          Please note: We are reviewing Western blot images included in the antibody testing data in our catalog, including those provided by third parties. Unless expressly labeled or annotated as “raw-unedited”, Western blot images included in the antibody testing data in our catalog may have been edited, optimized or otherwise adjusted for presentation.

          Product Details

          PA5-34794

          Applications
          Tested Dilution
          Publications

          Western Blot (WB)

          1:500-1:3,000
          -

          Immunohistochemistry (Paraffin) (IHC (P))

          1:100-1:1,000
          -
          Product Specifications

          Species Reactivity

          Human, Mouse

          Host/Isotype

          Rabbit / IgG

          Class

          Polyclonal

          Type

          Antibody

          Immunogen

          Recombinant protein encompassing a sequence within the center region of human TIP60. The exact sequence is proprietary.

          Conjugate

          Unconjugated Unconjugated Unconjugated

          Form

          Liquid

          Concentration

          1.67 mg/mL

          Amount

          167 µg

          Purification

          Antigen affinity chromatography

          Storage buffer

          PBS, pH 7, with 20% glycerol

          Contains

          0.025% ProClin 300

          Storage conditions

          Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.

          Shipping conditions

          Ambient (domestic); Wet ice (international)

          RRID

          AB_2552146

          Product Specific Information

          Recommended positive controls: Jurkat, Jurkat nucleus.

          Predicted reactivity: Mouse (99%), Rat (99%), Pig (99%), Bovine (98%).

          Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.

          Target Information

          TIP60 is a transcription factor, originally isolated as an HIV-1 TAT-interactive protein, known to interact with the viral TAT protein that leads to HTATIP polyubiquitination and targets it for degradation. It belongs to the MYST family of histone acetyl transferases (HATs). TIP60 is a catalytic subunit of the NuA4 histone acetyltransferase complex which is involved in transcriptional activation of select genes principally by acetylation of nucleosomal histone H4 and H2A. A co-regulator of transcription factors, TIP60 either promotes or suppresses tumorigenesis. TIP60 is an enzyme that has a role in DNA repair and apoptosis and is thought to play an important role in signal transduction.

          For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.

          References (0)

          Have you cited this product in a publication?
          Let us know so we can reference it here.
          Cite this product

          Bioinformatics

          Protein Aliases: 60 kDa Tat-interactive protein; cPLA(2)-interacting protein; cPLA2 interacting protein; histone acetyl transferase; histone acetyltransferase HTATIP; HIV-1 tat interactive protein 1, 60 kDa homolog; HIV-1 tat interactive protein 1, homolog; HIV-1 Tat interactive protein, 60kDa; K(lysine) acetyltransferase 5; K-acetyltransferase 5; lysine acetyltransferase 5; protein 2-hydroxyisobutyryltransferase KAT5; protein acetyltransferase KAT5; protein crotonyltransferase KAT5; protein lactyltransferase KAT5; Tat interacting protein, 60kDa; Tat-interactive protein-60; Tip60; Tip60 alpha; Tip60-A; Tip60 beta; Tip60-B; TIP60(HTATIP); unnamed protein product

          View more View less

          Gene Aliases: CPLA2; ESA1; HTATIP; HTATIP1; NEDFASB; PLIP; TIP; Tip55; TIP60; ZC2HC5

          View more View less

          UniProt ID: (Human) Q92993, (Mouse) Q8CHK4

          View more View less

          Entrez Gene ID: (Human) 10524, (Mouse) 81601

          View more View less

          Function(s)
          chromatin binding transcription coactivator activity histone acetyltransferase activity protein binding H4 histone acetyltransferase activity acetyltransferase activity H2A histone acetyltransferase activity histone H2AK5 acetyltransferase activity histone acetyltransferase activity (H4-K16 specific) peptide-lysine-N-acetyltransferase activity peptide 2-hydroxyisobutyryltransferase activity peptide lactyltransferase (CoA-dependent) activity peptide crotonyltransferase activity peptide butyryltransferase activity DNA-binding transcription factor binding phospholipase binding macromolecular complex binding
          Process(es)
          negative regulation of transcription from RNA polymerase II promoter establishment of mitotic spindle orientation double-strand break repair via homologous recombination DNA double-strand break processing nucleotide-excision repair double-strand break repair double-strand break repair via nonhomologous end joining regulation of transcription, DNA-templated regulation of transcription from RNA polymerase II promoter protein acetylation phosphatidylethanolamine biosynthetic process phosphatidylcholine biosynthetic process apoptotic process cellular response to DNA damage stimulus germ cell development spermatid development response to ionizing radiation positive regulation of autophagy positive regulation of triglyceride biosynthetic process peptidyl-lysine acetylation triglyceride biosynthetic process neural tube development neurogenesis DNA damage response, signal transduction by p53 class mediator negative regulation of interleukin-2 production cellular response to stress sperm chromatin condensation aggrephagy cellular response to glucose starvation positive regulation of circadian rhythm regulation of apoptotic process proteasome-mediated ubiquitin-dependent protein catabolic process positive regulation of innate immune response positive regulation of regulatory T cell differentiation negative regulation of myoblast differentiation positive regulation of myoblast differentiation negative regulation of transcription, DNA-templated positive regulation of transcription, DNA-templated positive regulation of transcription from RNA polymerase II promoter attachment of mitotic spindle microtubules to kinetochore regulation of cell cycle positive regulation of mitotic sister chromatid segregation protein targeting to vacuole involved in autophagy cellular response to glucose stimulus cellular response to estradiol stimulus membraneless organelle assembly DNA repair-dependent chromatin remodeling cGAS/STING signaling pathway cellular response to oxygen-containing compound positive regulation of signal transduction by p53 class mediator positive regulation of protein acetylation regulation of hematopoietic stem cell differentiation regulation of chromatin organization positive regulation of attachment of mitotic spindle microtubules to kinetochore positive regulation of double-strand break repair via homologous recombination positive regulation of aggrephagy lipid droplet disassembly protein localization to site of double-strand break negative regulation of double-strand break repair via homologous recombination regulation of double-strand break repair regulation of autophagy cellular senescence
          It has to be done as per old AB suggested Products section.

          Disclaimer

          Close

          Clicking the images or links will redirect you to a website hosted by BenchSci that provides third-party scientific content. Neither the content nor the BenchSci technology and processes for selection have been evaluated by us; we are providing them as-is and without warranty of any kind, including for use or application of the Thermo Fisher Scientific products presented.

          guarantee_icon

          Performance Guarantee

          If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*

          Learn more
          help_icon

          We're here to help

          Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.

          Contact tech support
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Responsibility Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          México flag icon
          México

          Your items have has been added!


          Host server : magellan-search-blue-6b866bbc8c-8lbpj:80/100.66.72.150:80.
          git-commit: 9d76681b284ad04e8ddcd97b79462898b32e8e5b
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.61.1-2026.10.99-1.0