Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • New Products
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          • Primary Antibodies ›
          • mCherry Antibodies

          Invitrogen

          mCherry Polyclonal Antibody

          32 Published Figures
          95 References
          View all (35) mCherry antibodies

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}

          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Tech Support
          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Tech Support
          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Tech Support

          Cite mCherry Polyclonal Antibody

          {{$ctrl.translations['antibody.pdp.skuNotes.additionalInformation'] || 'Additional Information'}}: {{$ctrl.translations['antibody.pdp.skuNotes.additionalInformation']}}:

          {{$ctrl.translations[$ctrl.isSkuNotesCollapsed ? 'antibody.pdp.skuNotes.viewMore' : 'antibody.pdp.skuNotes.viewLess']}}
          • Antibody Testing Data (8)
          • Published Figures (32)
          mCherry Antibody in Immunocytochemistry (ICC/IF)
          Group 53 Created with Sketch.
          mCherry Antibody in Immunocytochemistry (ICC/IF)
          Group 53 Created with Sketch.

          FIGURE: 1 / 40

          {{ $ctrl.currentElement.advancedVerification.fullName }}
          {{ $ctrl.currentElement.advancedVerification.text }}

          mCherry Antibody (PA5-34974) in ICC/IF

          mCherry Polyclonal Antibody detects mCherry protein at mCherry fusion protein by immunofluorescent analysis. Sample: mCherry fusion protein-transfected HeLa cells were fixed in 4% paraformaldehyde at RT for 30 min, then Triton X-100 for 1 min. Green: mCherry protein stained by mCherry Polyclonal Antibody (Product # PA5-34974) diluted at 1:500. Red: mCherry is expressed in the transfected cell . Blue: Hoechst 33342 staining. {{ $ctrl.currentElement.advancedVerification.fullName }} validation info. View more
          Published figure supplied by benchsci-logo
          PMID: {{$ctrl.currentElement.benchSciPubmedId}}
          View Product

          {{$ctrl.videoDesc}}

          {{$ctrl.videoLongDesc}}

          mCherry Antibody in Immunocytochemistry (ICC/IF)
          mCherry Antibody in Immunocytochemistry (ICC/IF)
          mCherry Antibody in Immunocytochemistry (ICC/IF)
          mCherry Antibody in Immunohistochemistry (IHC)
          mCherry Antibody in Western Blot (WB)
          mCherry Antibody in Western Blot (WB)
          mCherry Antibody in Western Blot (WB)
          mCherry Antibody in Immunoprecipitation (IP)
          mCherry Antibody in Immunocytochemistry (ICC/IF)
          mCherry Antibody in Immunocytochemistry (ICC/IF)
          mCherry Antibody in Western Blot, Immunocytochemistry (WB, ICC/IF)
          mCherry Antibody in Immunohistochemistry (Frozen) (IHC (F))
          mCherry Antibody in Immunohistochemistry, Immunohistochemistry (PFA fixed) (IHC, IHC (PFA))
          mCherry Antibody in Immunohistochemistry (IHC)
          mCherry Antibody in Immunohistochemistry (IHC)
          mCherry Antibody in Immunohistochemistry (IHC)
          mCherry Antibody in Immunohistochemistry (IHC)
          mCherry Antibody in Immunohistochemistry (IHC)
          mCherry Antibody in Immunohistochemistry (IHC)
          mCherry Antibody in Western Blot (WB)
          mCherry Antibody in Western Blot (WB)
          mCherry Antibody in Western Blot (WB)
          mCherry Antibody in Western Blot (WB)
          mCherry Antibody in Western Blot (WB)
          mCherry Antibody in Western Blot, Immunoprecipitation (WB, IP)
          mCherry Antibody in Western Blot (WB)
          mCherry Antibody in Western Blot (WB)
          mCherry Antibody in Western Blot (WB)
          mCherry Antibody in Western Blot (WB)
          mCherry Antibody in Western Blot (WB)
          mCherry Antibody in Western Blot (WB)
          mCherry Antibody in Western Blot (WB)
          mCherry Antibody in Western Blot (WB)
          mCherry Antibody in Western Blot (WB)
          mCherry Antibody in Western Blot (WB)
          mCherry Antibody in Western Blot (WB)
          mCherry Antibody in Western Blot (WB)
          mCherry Antibody in Western Blot (WB)
          mCherry Antibody in Immunoprecipitation (IP)
          mCherry Antibody in ChIP Assay (ChIP)

          Please note: We are reviewing Western blot images included in the antibody testing data in our catalog, including those provided by third parties. Unless expressly labeled or annotated as “raw-unedited”, Western blot images included in the antibody testing data in our catalog may have been edited, optimized or otherwise adjusted for presentation.

          Product Details

          PA5-34974

          Applications
          Tested Dilution
          Publications

          Western Blot (WB)

          1:1,000-1:10,000
          View 33 publications 33 publications

          Immunohistochemistry (IHC)

          -
          View 30 publications 30 publications

          Immunohistochemistry (Paraffin) (IHC (P))

          -
          View 1 publication 1 publication

          Immunohistochemistry (PFA fixed) (IHC (PFA))

          -
          View 1 publication 1 publication

          Immunohistochemistry (Frozen) (IHC (F))

          Assay-dependent
          View 3 publications 3 publications

          Immunocytochemistry (ICC/IF)

          1:100-1:1,000
          View 13 publications 13 publications

          Flow Cytometry (Flow)

          -
          View 2 publications 2 publications

          ELISA (ELISA)

          -
          View 1 publication 1 publication

          Immunoprecipitation (IP)

          1:100-1:500
          View 6 publications 6 publications

          ChIP assay (ChIP)

          -
          View 3 publications 3 publications

          Proximity Ligation Assay (PLA) (PLA)

          Assay-dependent
          -

          Mass Spectrometry (MS)

          -
          View 1 publication 1 publication

          Miscellaneous PubMed (Misc)

          -
          View 1 publication 1 publication
          Product Specifications

          Species Reactivity

          Tag

          Published species

          Tag

          Host/Isotype

          Rabbit / IgG

          Class

          Polyclonal

          Type

          Antibody

          Immunogen

          Full length mCherry recombinant protein.

          Conjugate

          Unconjugated Unconjugated Unconjugated

          Form

          Liquid

          Concentration

          0.22 mg/mL

          Amount

          22 µg

          Purification

          Antigen affinity chromatography

          Storage buffer

          PBS, pH 7, with 20% glycerol, 1% BSA

          Contains

          0.025% ProClin 300

          Storage conditions

          Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.

          Shipping conditions

          Ambient (domestic); Wet ice (international)

          RRID

          AB_2552323

          Product Specific Information

          Recommended positive controls: 293T whole cell extract, human PLK4-mCherry-transfected 293T.

          Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.

          Target Information

          MCherry is a fluorophore (a fluorescent molecule) used in biotechnology as a tracer to follow the flow of fluids, as a marker when tagged to molecules and cells components. mCherry is a monomeric fluorescent construct which absorbs at 587 nm and emits at 610 nm. It is resistant to photobleaching and is stable. It matures quickly, with a t 0. 5 of 15 minutes, allowing it to be visualized soon after translation.

          For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.

          Bioinformatics

          Protein Aliases: mCherry

          View more View less

          It has to be done as per old AB suggested Products section.

          Disclaimer

          Close

          Clicking the images or links will redirect you to a website hosted by BenchSci that provides third-party scientific content. Neither the content nor the BenchSci technology and processes for selection have been evaluated by us; we are providing them as-is and without warranty of any kind, including for use or application of the Thermo Fisher Scientific products presented.

          guarantee_icon

          Performance Guarantee

          If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*

          Learn more
          help_icon

          We're here to help

          Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.

          Contact tech support
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Responsibility Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          México flag icon
          México

          Your items have has been added!


          Host server : magellan-search-blue-6b866bbc8c-vnqbn:80/100.66.76.64:80.
          git-commit: 9d76681b284ad04e8ddcd97b79462898b32e8e5b
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.61.1-2026.10.99-1.0