Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • New Products
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          • Primary Antibodies ›
          • TGFBR2 Antibodies

          Invitrogen

          TGFBR2 Polyclonal Antibody

          Advanced Verification
          1 Published Figure
          7 References
          View all (27) TGFBR2 antibodies

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}

          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Tech Support
          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Tech Support
          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Tech Support

          Cite TGFBR2 Polyclonal Antibody

          {{$ctrl.translations['antibody.pdp.skuNotes.additionalInformation'] || 'Additional Information'}}: {{$ctrl.translations['antibody.pdp.skuNotes.additionalInformation']}}:

          {{$ctrl.translations[$ctrl.isSkuNotesCollapsed ? 'antibody.pdp.skuNotes.viewMore' : 'antibody.pdp.skuNotes.viewLess']}}
          • Antibody Testing Data (8)
          • Published Figures (1)
          • Advanced Verification (1)
          TGFBR2 Antibody in Immunocytochemistry (ICC/IF)
          Group 53 Created with Sketch.
          TGFBR2 Antibody in Immunocytochemistry (ICC/IF)
          Group 53 Created with Sketch.

          FIGURE: 1 / 10

          {{ $ctrl.currentElement.advancedVerification.fullName }}
          {{ $ctrl.currentElement.advancedVerification.text }}

          TGFBR2 Antibody (PA5-35076) in ICC/IF

          Immunofluorescence analysis of TGFBR2 was performed using 70% confluent log phase HT-29 cells. The cells were fixed with 4% paraformaldehyde for 10 minutes, permeabilized with 0.1% Triton™ X-100 for 15 minutes, and blocked with 1% BSA for 1 hour at room temperature. The cells were labeled with TGFBR2 Rabbit Polyclonal Antibody(Product # PA5-35076) at 1:100 dilution in 0.1% BSA, incubated at 4 degree Celsius overnight and then labeled with Goat anti-Rabbit IgG (Heavy Chain) Superclonal™ Secondary Antibody, Alexa Fluor® 488 conjugate (Produc... View More {{ $ctrl.currentElement.advancedVerification.fullName }} validation info. View more
          Published figure supplied by benchsci-logo
          PMID: {{$ctrl.currentElement.benchSciPubmedId}}
          View Product

          {{$ctrl.videoDesc}}

          {{$ctrl.videoLongDesc}}

          TGFBR2 Antibody in Immunocytochemistry (ICC/IF)
          TGFBR2 Antibody in Immunocytochemistry (ICC/IF)
          TGFBR2 Antibody in Western Blot (WB)
          TGFBR2 Antibody in Western Blot (WB)
          TGFBR2 Antibody in Western Blot (WB)
          TGFBR2 Antibody in Western Blot (WB)
          TGFBR2 Antibody in Western Blot (WB)
          TGFBR2 Antibody in Western Blot (WB)
          TGFBR2 Antibody in Immunocytochemistry (ICC/IF)
          TGFBR2 Antibody

          Please note: We are reviewing Western blot images included in the antibody testing data in our catalog, including those provided by third parties. Unless expressly labeled or annotated as “raw-unedited”, Western blot images included in the antibody testing data in our catalog may have been edited, optimized or otherwise adjusted for presentation.

          TGFBR2 Polyclonal Antibody

          Product Details

          PA5-35076

          Applications
          Tested Dilution
          Publications

          Western Blot (WB)

          1:1,000
          View 3 publications 3 publications

          Immunohistochemistry (IHC)

          -
          View 3 publications 3 publications

          Immunohistochemistry (Paraffin) (IHC (P))

          1:25
          -

          Immunocytochemistry (ICC/IF)

          1:10-1:50
          View 1 publication 1 publication
          Product Specifications

          Species Reactivity

          Human, Mouse

          Published species

          Human, Mouse

          Host/Isotype

          Rabbit / IgG

          Class

          Polyclonal

          Type

          Antibody

          Immunogen

          KLH conjugated synthetic peptide between 13-40 amino acids from the N-terminal region of human TGFBR2
          3D Epitope / Immunogen

          Conjugate

          Unconjugated Unconjugated Unconjugated

          Form

          Liquid

          Concentration

          0.5 mg/mL

          Amount

          200 µg

          Purification

          Protein A, Antigen affinity chromatography

          Storage buffer

          PBS, pH 7.4

          Contains

          0.09% sodium azide

          Storage conditions

          Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.

          Shipping conditions

          Ambient (domestic); Wet ice (international)

          RRID

          AB_2552386

          Target Information

          Transforming Growth Factor, beta receptor 2 (AAT3, FAA3, MFS2, RIIC, LDS1B, LDS2B, TAAD2, TGFR-2, TGFbeta-RII) is a member of the TGF-beta family of receptor Serine/Threonine kinases. It is involved in the regulation of cell proliferation. Mitochondrial ATP synthase catalyzes ATP synthesis, utilizing an electrochemical gradient of protons across the inner membrane during oxidative phosphorylation. ATP synthase is composed of two linked multi-subunit complexes: the soluble catalytic core, F1, and the membrane-spanning component, Fo, comprising the proton channel. The catalytic portion of mitochondrial ATP synthase consists of 5 different subunits (alpha, beta, gamma, delta, and epsilon) assembled with a stoichiometry of 3 alpha, 3 beta, and a single representative of the other 3. The proton channel consists of three main subunits (a, b, c). TGFBR2 encodes the gamma subunit of the catalytic core. Alternatively spliced transcript variants encoding different isoforms have been identified. TGFBR2 also has a pseudogene on chromosome 14.

          For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.

          Bioinformatics

          Protein Aliases: Ser/Thr protein kinase; tgf beta receptor 2; TGF-beta receptor II; TGF-beta receptor type II; TGF-beta receptor type IIB; TGF-beta type II receptor; TGFbeta RII; TGFBR; TGFBR2-SE; TGFR-2; TGFR2; transforming growth factor beta receptor II; transforming growth factor beta receptor II (70/80kDa) isoform TGFBR2_1; transforming growth factor beta receptor II (70/80kDa) isoform TGFBR2_2; transforming growth factor beta receptor type IIC; transforming growth factor, beta receptor II (70/80kDa); transforming growth factor, beta receptor II alpha; transforming growth factor, beta receptor II beta; transforming growth factor, beta receptor II delta; transforming growth factor, beta receptor II epsilon; transforming growth factor, beta receptor II gamma; transforming growth factor-beta receptor type II; unnamed protein product

          View more View less

          Gene Aliases: 1110020H15Rik; AAT3; DNIIR; FAA3; LDS1B; LDS2; LDS2B; MFS2; RIIC; RIIDN; TAAD2; TbetaR-II; TbetaRII; TBR-II; TBRII; TGFbeta-RII; TGFR-2

          View more View less

          UniProt ID: (Human) P37173, (Mouse) Q62312

          View more View less

          Entrez Gene ID: (Human) 7048, (Mouse) 21813

          View more View less

          Function(s)
          protein kinase activity protein serine/threonine kinase activity transmembrane receptor protein serine/threonine kinase activity transforming growth factor beta-activated receptor activity transforming growth factor beta receptor activity, type II protein binding ATP binding glycosaminoglycan binding activin receptor activity, type I kinase activator activity type I transforming growth factor beta receptor binding type III transforming growth factor beta receptor binding signaling receptor activity SMAD binding metal ion binding activin binding transforming growth factor beta binding binding, bridging mitogen-activated protein kinase kinase kinase binding
          Process(es)
          blood vessel development patterning of blood vessels vasculogenesis epithelial to mesenchymal transition heart looping positive regulation of mesenchymal cell proliferation positive regulation of tolerance induction to self antigen positive regulation of B cell tolerance induction positive regulation of T cell tolerance induction outflow tract septum morphogenesis membranous septum morphogenesis outflow tract morphogenesis aortic valve morphogenesis atrioventricular valve morphogenesis tricuspid valve morphogenesis cardiac left ventricle morphogenesis endocardial cushion fusion apoptotic process transmembrane receptor protein serine/threonine kinase signaling pathway transforming growth factor beta receptor signaling pathway nervous system development brain development heart development positive regulation of cell proliferation response to xenobiotic stimulus positive regulation of epithelial to mesenchymal transition cell differentiation activin receptor signaling pathway response to lipid embryonic hemopoiesis aorta morphogenesis regulation of cell proliferation myeloid dendritic cell differentiation animal organ development blood vessel morphogenesis embryonic cranial skeleton morphogenesis artery morphogenesis positive regulation of developmental process positive regulation of NK T cell differentiation cartilage development regulation of multicellular organismal process palate development positive regulation of SMAD protein import into nucleus ventricular septum morphogenesis secondary palate development response to cholesterol cellular response to growth factor stimulus positive regulation of epithelial to mesenchymal transition involved in endocardial cushion formation cell proliferation involved in endocardial cushion morphogenesis superior endocardial cushion morphogenesis inferior endocardial cushion morphogenesis miRNA transport positive regulation of reactive oxygen species metabolic process positive regulation of CD4-positive, alpha-beta T cell proliferation regulation of stem cell differentiation in utero embryonic development lens development in camera-type eye growth plate cartilage development growth plate cartilage chondrocyte growth protein phosphorylation receptor-mediated endocytosis Notch signaling pathway smoothened signaling pathway gastrulation negative regulation of cell proliferation organ morphogenesis regulation of gene expression positive regulation of epithelial cell migration lung development positive regulation of angiogenesis positive regulation of smooth muscle cell proliferation negative regulation of cardiac muscle cell proliferation SMAD protein signal transduction lung morphogenesis bronchus development bronchus morphogenesis trachea morphogenesis trachea formation mammary gland morphogenesis lung lobe morphogenesis lens fiber cell apoptotic process
          It has to be done as per old AB suggested Products section.

          Disclaimer

          Close

          Clicking the images or links will redirect you to a website hosted by BenchSci that provides third-party scientific content. Neither the content nor the BenchSci technology and processes for selection have been evaluated by us; we are providing them as-is and without warranty of any kind, including for use or application of the Thermo Fisher Scientific products presented.

          guarantee_icon

          Performance Guarantee

          If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*

          Learn more
          help_icon

          We're here to help

          Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.

          Contact tech support
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Responsibility Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          México flag icon
          México

          Your items have has been added!


          Host server : magellan-search-blue-6b866bbc8c-mwps6:80/100.66.76.64:80.
          git-commit: 9d76681b284ad04e8ddcd97b79462898b32e8e5b
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.61.1-2026.10.99-1.0