Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • New Products
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          • Primary Antibodies ›
          • SLC11A2 Antibodies

          Invitrogen

          SLC11A2 Polyclonal Antibody

          3 Published Figures
          3 References
          View all (18) SLC11A2 antibodies

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}

          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Datasheet
          Certificates
          Protocols
          Questions & Answers

          Cite SLC11A2 Polyclonal Antibody

          Additional Information:

          {{$ctrl.isSkuNotesCollapsed ? 'View more' : 'View less'}}
          • Antibody Testing Data (5)
          • Published Figures (3)
          SLC11A2 Antibody in Immunocytochemistry (ICC/IF)
          Group 53 Created with Sketch.
          SLC11A2 Antibody in Immunocytochemistry (ICC/IF)
          Group 53 Created with Sketch.

          FIGURE: 1 / 8

          {{ $ctrl.currentElement.advancedVerification.fullName }}
          {{ $ctrl.currentElement.advancedVerification.text }}

          SLC11A2 Antibody (PA5-35136) in ICC/IF

          Immunofluorescence analysis of SLC11A2 was performed using 70% confluent log phase Hep G2 cells. The cells were fixed with 4% paraformaldehyde for 10 minutes, permeabilized with 0.1% Triton™ X-100 for 15 minutes, and blocked with 1% BSA for 1 hour at room temperature. The cells were labeled with SLC11A2 Polyclonal Antibody (Product # PA5-35136) at 1:50 dilution in 0.1% BSA, incubated at 4 degree Celsius overnight and then labeled with Goat anti-Rabbit IgG (Heavy Chain) Superclonal™ Secondary Antibody, Alexa Fluor® 488 conjugate (Product # ... View More {{ $ctrl.currentElement.advancedVerification.fullName }} validation info. View more
          Published figure supplied by benchsci-logo
          PMID: {{$ctrl.currentElement.benchSciPubmedId}}
          View Product

          {{$ctrl.videoDesc}}

          {{$ctrl.videoLongDesc}}

          SLC11A2 Antibody in Immunocytochemistry (ICC/IF)
          SLC11A2 Antibody in Immunocytochemistry (ICC/IF)
          SLC11A2 Antibody in Western Blot (WB)
          SLC11A2 Antibody in Western Blot (WB)
          SLC11A2 Antibody in Western Blot (WB)
          SLC11A2 Antibody in Western Blot (WB)
          SLC11A2 Antibody in Western Blot (WB)
          SLC11A2 Antibody in Western Blot (WB)

          Please note: We are reviewing Western blot images included in the antibody testing data in our catalog, including those provided by third parties. Unless expressly labeled or annotated as “raw-unedited”, Western blot images included in the antibody testing data in our catalog may have been edited, optimized or otherwise adjusted for presentation.

          SLC11A2 Polyclonal Antibody

          Product Details

          PA5-35136

          Applications
          Tested Dilution
          Publications

          Western Blot (WB)

          1:2,000
          View 3 publications 3 publications

          Immunocytochemistry (ICC/IF)

          1:50-1:200
          -
          Product Specifications

          Species Reactivity

          Human, Mouse, Rat

          Published species

          Human, Mouse

          Host/Isotype

          Rabbit / IgG

          Class

          Polyclonal

          Type

          Antibody

          Immunogen

          KLH conjugated synthetic peptide between 262-291 amino acids from the central region of human SLC11A2
          3D Epitope / Immunogen

          Conjugate

          Unconjugated Unconjugated Unconjugated

          Form

          Liquid

          Concentration

          0.5 mg/mL

          Amount

          200 µg

          Purification

          Protein A, Antigen affinity chromatography

          Storage buffer

          PBS, pH 7.4

          Contains

          0.09% sodium azide

          Storage conditions

          Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.

          Shipping conditions

          Ambient (domestic); Wet ice (international)

          RRID

          AB_2552446

          Target Information

          This gene encodes a member of the solute carrier family 11 protein family. The product of this gene transports divalent metals and is involved in iron absorption. Mutations in this gene are associated with hypochromic microcytic anemia with iron overload. A related solute carrier family 11 protein gene is located on chromosome 2. Multiple transcript variants encoding different isoforms have been found for this gene.

          For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.

          Bioinformatics

          Protein Aliases: Dct-1; DCT1; divalent cation transporter 1; divalent metal transporter 1; DMT-1; microcytic anemia, viable anaemia; natural resistance-associated macrophage protein 2; NRAMP 2; Slc11a2; solute carrier family 11 (proton-coupled divalent metal ion transporter), member 2; solute carrier family 11 (proton-coupled divalent metal ion transporters), member 2; solute carrier family 11 member 2; Solute carrier family 11 member 2 (natural resistance-associated macrophage protein 2); unnamed protein product

          View more View less

          Gene Aliases: AHMIO1; DCT1; DMT1; mk; NRAMP2; van

          View more View less

          UniProt ID: (Human) P49281, (Mouse) P49282, (Rat) O54902

          View more View less

          Entrez Gene ID: (Human) 4891, (Mouse) 18174, (Rat) 25715

          View more View less

          Function(s)
          copper ion transmembrane transporter activity iron ion transmembrane transporter activity manganese ion transmembrane transporter activity zinc ion transmembrane transporter activity protein binding calcium ion transmembrane transporter activity cadmium ion transmembrane transporter activity cobalt ion transmembrane transporter activity ferrous iron transmembrane transporter activity lead ion transmembrane transporter activity nickel cation transmembrane transporter activity vanadium ion transmembrane transporter activity solute:proton symporter activity transmembrane transporter activity cadmium ion binding metal ion transmembrane transporter activity transition metal ion transmembrane transporter activity chromium ion transmembrane transporter activity retromer complex binding iron ion binding copper ion binding zinc ion binding hydrogen ion transmembrane transporter activity nickel cation binding manganese ion binding cobalt ion binding
          Process(es)
          transition metal ion transport porphyrin-containing compound metabolic process porphyrin-containing compound biosynthetic process heme biosynthetic process cobalt ion transport copper ion transport iron ion transport manganese ion transport cellular iron ion homeostasis learning or memory response to iron ion response to manganese ion nickel cation transport vanadium ion transport lead ion transport metal ion transport cellular manganese ion homeostasis iron assimilation cellular response to oxidative stress iron ion transmembrane transport nickel cation transmembrane transport dendrite morphogenesis erythrocyte development multicellular organismal iron ion homeostasis cadmium ion transmembrane transport manganese ion transmembrane transport zinc II ion transmembrane transport copper ion import across plasma membrane hydrogen ion transmembrane transport response to hypoxia detection of oxygen response to lead ion response to cadmium ion cellular response to iron ion cellular response to tumor necrosis factor cellular response to hypoxia copper ion transmembrane transport transmembrane transport
          It has to be done as per old AB suggested Products section.

          Disclaimer

          Close

          Clicking the images or links will redirect you to a website hosted by BenchSci that provides third-party scientific content. Neither the content nor the BenchSci technology and processes for selection have been evaluated by us; we are providing them as-is and without warranty of any kind, including for use or application of the Thermo Fisher Scientific products presented.

          guarantee_icon

          Performance Guarantee

          If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*

          Learn more
          help_icon

          We're here to help

          Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.

          Contact tech support
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-blue-5bc99cc877-bn8h2:80/100.66.76.64:80.
          git-commit: a70b05477dc28cec55d7eafa2ce360191b99e591
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.58.0-2026.09.99-1.0