Search
Search
Invitrogen
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}
Please note: We are reviewing Western blot images included in the antibody testing data in our catalog, including those provided by third parties. Unless expressly labeled or annotated as “raw-unedited”, Western blot images included in the antibody testing data in our catalog may have been edited, optimized or otherwise adjusted for presentation.
This antibody detects endogenous protein at a molecular weight of 47 kDa.
Purity is >95% by SDS-PAGE.
Sox genes comprise a family of genes that are related to the mammalian sex determining gene SRY. These genes similarly contain sequences that encode for the HMG-box domain, which is responsible for the sequence-specific DNA-binding activity. Sox genes encode putative transcriptional regulators implicated in the decision of cell fates during development and the control of diverse developmental processes. The highly complex group of Sox genes cluster at least 40 different loci that rapidly diverged in various animal lineages. At present, 30 Sox genes have been identified. Members of this family have been shown to be conserved during evolution and to play key roles during animal development. Some are involved in human diseases, including sex reversal.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: mSox9; ragged; SRY (sex determining region Y)-box 9; SRY (sex determining region Y)-box 9 (campomelic dysplasia, autosomal sex-reversal); SRY-box containing gene 17; SRY-box containing gene 18; SRY-box containing gene 8; SRY-box containing gene 9; Sry-related HMG-box gene 18; Transcription factor SOX-17; Transcription factor SOX-18; Transcription factor SOX-8; Transcription factor SOX-9
Gene Aliases: 2010306G03Rik; AI385749; AV220920; mKIAA4243; Ra; Ragl; Sox-17; Sox-18; Sox-8; Sox-9; Sox17; Sox18; Sox8; Sox9
UniProt ID: (Mouse) Q61473, (Mouse) P43680, (Mouse) Q04886, (Rat) F1LYL9, (Mouse) Q04887
Entrez Gene ID: (Mouse) 20671, (Rat) 312936, (Rat) 311723, (Mouse) 20672, (Mouse) 20681, (Rat) 302993, (Rat) 140586, (Mouse) 20682
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn more
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support