Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          • Primary Antibodies ›
          • FOXO1/FOXO3 Antibodies

          Invitrogen

          Phospho-FOXO1/FOXO3 (Ser322, Ser325) Polyclonal Antibody

          View all (2) FOXO1/FOXO3 antibodies

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}

          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Tech Support
          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Tech Support
          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Tech Support

          Cite Phospho-FOXO1/FOXO3 (Ser322, Ser325) Polyclonal Antibody

          Additional Information:
          {{banner.description}}
          {{$ctrl.isSkuNotesCollapsed ? 'View more' : 'View less'}}
          • Antibody Testing Data (1)
          Phospho-FOXO1/FOXO3 (Ser322, Ser325) Antibody in Western Blot (WB)
          Group 53 Created with Sketch.
          Phospho-FOXO1/FOXO3 (Ser322, Ser325) Antibody in Western Blot (WB)
          Group 53 Created with Sketch.

          FIGURE: 1 / 1

          {{ $ctrl.currentElement.advancedVerification.fullName }}
          {{ $ctrl.currentElement.advancedVerification.text }}

          Phospho-FOXO1/FOXO3 (Ser322, Ser325) Antibody (PA5-40217) in WB

          Western Blot analysis of Phospho-FOXO1/FOXO3 (Ser322, Ser325) was performed using 3T3 cells. The membrane was probed with Phospho-FOXO1/FOXO3 (Ser322, Ser325) Polyclonal Antibody (Product # PA5-40217). {{ $ctrl.currentElement.advancedVerification.fullName }} validation info. View more
          Published figure supplied by benchsci-logo
          PMID: {{$ctrl.currentElement.benchSciPubmedId}}
          View Product

          {{$ctrl.videoDesc}}

          {{$ctrl.videoLongDesc}}

          Phospho-FOXO1/FOXO3 (Ser322, Ser325) Antibody in Western Blot (WB)

          Please note: We are reviewing Western blot images included in the antibody testing data in our catalog, including those provided by third parties. Unless expressly labeled or annotated as “raw-unedited”, Western blot images included in the antibody testing data in our catalog may have been edited, optimized or otherwise adjusted for presentation.

          Phospho-FOXO1/FOXO3 (Ser322, Ser325) Polyclonal Antibody

          Product Details

          PA5-40217

          Applications
          Tested Dilution
          Publications

          Western Blot (WB)

          1:500-1:2,000
          -

          Immunohistochemistry (IHC)

          1:100-1:300
          -

          ELISA (ELISA)

          1:5,000
          -
          Product Specifications

          Species Reactivity

          Human, Mouse, Rat

          Host/Isotype

          Rabbit / IgG

          Class

          Polyclonal

          Type

          Antibody

          Immunogen

          Synthesized peptide derived from human FoxO1/3 around the phosphorylation site of S322/S325.
          3D Epitope / Immunogen

          Conjugate

          Unconjugated Unconjugated Unconjugated

          Form

          Liquid

          Concentration

          1 mg/mL

          Amount

          100 µg

          Purification

          Antigen affinity chromatography

          Storage buffer

          PBS, pH 7.4, with 0.5% BSA, 50% glycerol

          Contains

          0.02% sodium azide

          Storage conditions

          -20°C, Avoid Freeze/Thaw Cycles

          Shipping conditions

          Ambient (domestic); Wet ice (international)

          RRID

          AB_2576790

          Product Specific Information

          This antibody is predicted to react with human and rat based on sequence homology.

          Target Information

          a transcription factor of the forkhead family. A central regulator of metabolism in several cell types. May play an important role in myogenic growth and differentiation, and in the regulation of lipolysis in adipocytes by controlling the expression of adipose triglyceride lipase (ATGL), the rate-limiting lipolytic enzyme. Translocation of this gene with PAX3 has been associated with alveolar rhabdomyosarcoma. Contains 1 fork-head domain.

          For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.

          References (0)

          Have you cited this product in a publication?
          Let us know so we can reference it here.
          Cite this product

          Bioinformatics

          Protein Aliases: AFX; AFX1; forkhead box O1a; forkhead box O1A (rhabdomyosarcoma); forkhead box O3A; Forkhead box protein O1A; forkhead homolog (rhabdomyosarcoma) like 1; Forkhead in rhabdomyosarcoma; forkhead in rhabdomyosarcoma-like 1; forkhead protein 1; forkhead protein FKHR2; forkhead, Drosophila, homolog of, in rhabdomyosarcoma; forkhead, Drosophila, homolog of, in rhabdomyosarcoma-like 1; FOXO3A-; FOXO4; member of FKHR-subclass forkhead/winged helix family; MGC120490; MLLT7; unnamed protein product

          View more View less

          Gene Aliases: 1110048B16Rik; 2010203A17Rik; AF6q21; Afxh; FKH1; FKHR; Fkhr1; Fkhr2; FKHRL1; FKHRL1P2; FOXO1A; FOXO2; FOXO3A

          View more View less

          UniProt ID: (Human) Q12778, (Human) O43524, (Rat) G3V7R4, (Mouse) Q9R1E0, (Mouse) Q9WVH4

          View more View less

          Entrez Gene ID: (Human) 2308, (Human) 2309, (Rat) 84482, (Mouse) 56458, (Mouse) 56484, (Rat) 294515

          View more View less

          Function(s)
          RNA polymerase II core promoter proximal region sequence-specific DNA binding RNA polymerase II transcription factor activity, sequence-specific DNA binding transcriptional repressor activity, RNA polymerase II transcription regulatory region sequence-specific binding transcriptional activator activity, RNA polymerase II transcription regulatory region sequence-specific binding nucleic acid binding DNA binding chromatin binding transcription factor activity, sequence-specific DNA binding protein binding beta-catenin binding chromatin DNA binding ubiquitin protein ligase binding sequence-specific DNA binding protein phosphatase 2A binding promoter-specific chromatin binding transcription regulatory region sequence-specific DNA binding transcription cofactor binding protein kinase binding mitochondrial RNA polymerase binding promoter specificity activity sequence-specific double-stranded DNA binding transcription coactivator binding transcription factor binding
          Process(es)
          negative regulation of transcription from RNA polymerase II promoter blood vessel development regulation of gluconeogenesis regulation of transcription, DNA-templated regulation of transcription from RNA polymerase II promoter apoptotic process cellular response to DNA damage stimulus insulin receptor signaling pathway cellular response to starvation gene expression positive regulation of autophagy negative regulation of gene expression peptidyl-serine phosphorylation peptidyl-threonine phosphorylation response to insulin cellular response to insulin stimulus negative regulation of stress-activated MAPK cascade positive regulation of smooth muscle cell apoptotic process cellular response to oxidative stress regulation of cell proliferation glucose homeostasis positive regulation of apoptotic process negative regulation of apoptotic process negative regulation of fat cell differentiation positive regulation of gluconeogenesis positive regulation of protein catabolic process positive regulation of transcription, DNA-templated positive regulation of transcription from RNA polymerase II promoter negative regulation of insulin secretion regulation of transcription initiation from RNA polymerase II promoter enamel mineralization cellular response to hydrogen peroxide response to fatty acid cellular response to hyperoxia cellular response to dexamethasone stimulus cellular response to nitric oxide negative regulation of canonical Wnt signaling pathway neuronal stem cell population maintenance response to fluoride negative regulation of cardiac muscle hypertrophy in response to stress positive regulation of complement-dependent cytotoxicity regulation of neural precursor cell proliferation regulation of reactive oxygen species metabolic process ovulation from ovarian follicle initiation of primordial ovarian follicle growth antral ovarian follicle growth oocyte maturation transcription from mitochondrial promoter regulation of translation phagocytosis response to xenobiotic stimulus positive regulation of muscle atrophy DNA damage response, signal transduction by p53 class mediator negative regulation of cell migration response to nutrient levels tumor necrosis factor-mediated signaling pathway cellular response to glucose starvation response to starvation positive regulation of neuron apoptotic process positive regulation of regulatory T cell differentiation positive regulation of erythrocyte differentiation negative regulation of neuron differentiation brain morphogenesis cellular response to glucose stimulus cellular response to corticosterone stimulus response to dexamethasone extrinsic apoptotic signaling pathway in absence of ligand positive regulation of hydrogen peroxide-mediated programmed cell death positive regulation of pri-miRNA transcription from RNA polymerase II promoter positive regulation of reactive oxygen species biosynthetic process cellular response to beta-amyloid cellular response to nerve growth factor stimulus response to water-immersion restraint stress positive regulation of endothelial cell apoptotic process temperature homeostasis cellular glucose homeostasis protein acetylation fat cell differentiation negative regulation of transcription, DNA-templated cellular response to cold energy homeostasis gluconeogenesis
          It has to be done as per old AB suggested Products section.

          Disclaimer

          Close

          Clicking the images or links will redirect you to a website hosted by BenchSci that provides third-party scientific content. Neither the content nor the BenchSci technology and processes for selection have been evaluated by us; we are providing them as-is and without warranty of any kind, including for use or application of the Thermo Fisher Scientific products presented.

          guarantee_icon

          Performance Guarantee

          If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*

          Learn more
          help_icon

          We're here to help

          Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.

          Contact tech support
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Responsibility Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          México flag icon
          México

          Your items have has been added!


          Host server : magellan-search-green-5799cd5976-swbc5:80/100.66.77.38:80.
          git-commit: e30ab741173a2eb62411af663260592d4bdb9634
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.55.0-2026.07.30-1.0