Search
Search
Invitrogen
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}
Peptide sequence: PEPPCCAAPD AAAAAFPPCA AAASPPLYSQ VPDRLVLPAT RPGPGPLPAE
Sequence homology: Human: 100%
Forkhead Box Protein E3 (FOXE3, forkhead-related protein FKHL12, forkhead-related transcription factor 8) is a forkhead/winged helix transcription factor, which is expressed in the developing lens from the start of lens placode induction and becomes restricted to the anterior proliferating cells when lens fiber differentiation begins.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Forkhead box protein E3; forkhead, drosophila, homolog-like 12; Forkhead-related protein FKHL12; Forkhead-related transcription factor 8; FREAC-8
Gene Aliases: AAT11; ASGD2; CATC3; CTRCT34; FKHL12; FOXE3; FREAC8
UniProt ID: (Human) Q13461
Entrez Gene ID: (Human) 2301
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn more
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support