Search
Search
Invitrogen
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}
Peptide sequence: FGNPNRPVAS TRLPSDTDDS DASEDPGPGA ERGGASSSCC EEEQTQGRGA
Sequence homology: Human: 100%
TSR3 is a probable pre-rRNA processing protein involved in ribosome biogenesis.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: 18S rRNA aminocarboxypropyltransferase; 20S S rRNA accumulation protein 3 homolog; HsTsr3; MGC24381; probable ribosome biogenesis protein C16orf42; ribosome biogenesis protein TSR3 homolog; TSR3, 20S rRNA accumulation, homolog; TSR3, acp transferase ribosome maturation factor
Gene Aliases: C16orf42; HsTsr3; TSR3; UND313L
UniProt ID: (Human) Q9UJK0
Entrez Gene ID: (Human) 115939
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn more
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support