Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • New Products
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          • Primary Antibodies ›
          • CCL2 (MCP-1) Antibodies

          Invitrogen

          MCP-1 Polyclonal Antibody

          Advanced Verification
          1 Reference
          View all (62) CCL2 (MCP-1) antibodies

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}

          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Tech Support
          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Tech Support
          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Tech Support

          Cite MCP-1 Polyclonal Antibody

          {{$ctrl.translations['antibody.pdp.skuNotes.additionalInformation'] || 'Additional Information'}}: {{$ctrl.translations['antibody.pdp.skuNotes.additionalInformation']}}:

          {{$ctrl.translations[$ctrl.isSkuNotesCollapsed ? 'antibody.pdp.skuNotes.viewMore' : 'antibody.pdp.skuNotes.viewLess']}}
          • Antibody Testing Data (2)
          • Advanced Verification (1)
          MCP-1 Antibody in Western Blot (WB)
          Group 53 Created with Sketch.
          MCP-1 Antibody in Western Blot (WB)
          Group 53 Created with Sketch.

          FIGURE: 1 / 3

          {{ $ctrl.currentElement.advancedVerification.fullName }}
          {{ $ctrl.currentElement.advancedVerification.text }}

          MCP-1 Antibody (PA5-46954) in WB

          Western blot analysis of MCP-1 in 25 ng recombinant Mouse CCL2/JE/MCP‚1, recombinant Human CCL2/JE/MCP‚1, recombinant Rat CCL2/JE/MCP‚1, recombinant Mouse CCL11/Eotaxin, recombinant Mouse CCL12/MCP‚5, and recombinant Mouse CCL7/MCP‚3/MARC. Samples were incubated in MCP-1 polyclonal antibody (Product # PA5-46954) using a dilution of 0.1 µg/mL followed by a HRP-conjugated Anti-Goat IgG secondary antibody. A specific band was detected for CCL2/JE/MCP‚1 at approximately 10 kDa (as indicated). This experiment was conducted under reducing condit... View More {{ $ctrl.currentElement.advancedVerification.fullName }} validation info. View more
          Published figure supplied by benchsci-logo
          PMID: {{$ctrl.currentElement.benchSciPubmedId}}
          View Product

          {{$ctrl.videoDesc}}

          {{$ctrl.videoLongDesc}}

          MCP-1 Antibody in Western Blot (WB)
          MCP-1 Antibody in Neutralization (Neu)
          MCP-1 Antibody

          Please note: We are reviewing Western blot images included in the antibody testing data in our catalog, including those provided by third parties. Unless expressly labeled or annotated as “raw-unedited”, Western blot images included in the antibody testing data in our catalog may have been edited, optimized or otherwise adjusted for presentation.

          MCP-1 Polyclonal Antibody

          Product Details

          PA5-46954

          Applications
          Tested Dilution
          Publications

          Western Blot (WB)

          0.1 µg/mL
          View 1 publication 1 publication

          ELISA (ELISA)

          0.1-0.4 µg/mL (Detection), 0.2-0.8 µg/mL (Capture)
          -

          Neutralization (Neu)

          0.5-2.5 µg/mL
          -
          Product Specifications

          Species Reactivity

          Mouse

          Published species

          Mouse

          Host/Isotype

          Goat / IgG

          Class

          Polyclonal

          Type

          Antibody

          Immunogen

          E. coli-derived recombinant mouse CCL2/JE/MCP-1 Gln24-Arg96
          3D Epitope / Immunogen

          Conjugate

          Unconjugated Unconjugated Unconjugated

          Form

          Lyophilized

          Concentration

          0.2 mg/mL once reconstituted

          Amount

          100 µg

          Purification

          Antigen affinity chromatography

          Storage buffer

          PBS with 5% trehalose

          Contains

          No Preservative

          Storage conditions

          -20°C, Avoid Freeze/Thaw Cycles

          Shipping conditions

          Ambient (domestic); Wet ice (international)

          RRID

          AB_2609536

          Product Specific Information

          In sandwich ELISAs, less than 0.01% cross-reactivity with recombinant human (rh) MCP-1, rhMCP-2, rhMCP-3, recombinant mouse (rm) MARC, rhMCP-4, rmMCP-5, rmMDC, rmMIP-1 alpha, rmMIP-1 beta, rmMIP-1 gamma, rmMIP-2, rmMIP-3 beta, rmRANTES, rmTARC, and rmTeck is observed.

          Reconstitute at 0.2 mg/mL in sterile PBS.

          Endoxin level is <0.10 EU per 1 µg of the antibody by the LAL method.

          Target Information

          This gene is one of several cytokine genes clustered on the q-arm of chromosome 17. Cytokines are a family of secreted proteins involved in immunoregulatory and inflammatory processes. The protein encoded by this gene is structurally related to the CXC subfamily of cytokines. Members of this subfamily are characterized by two cysteines separated by a single amino acid. This cytokine displays chemotactic activity for monocytes and basophils but not for neutrophils or eosinophils. It has been implicated in the pathogenesis of diseases characterized by monocytic infiltrates, like psoriasis, rheumatoid arthritis and atherosclerosis. It binds to chemokine receptors CCR2 and CCR4.

          For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.

          Bioinformatics

          Protein Aliases: Ccl2; chemokine (C-C motif) ligand 2; H-MCP-1; JE; MGC9434; monocyte chemoattractant protein 1; monocyte chemotactic protein 1; platelet-derived growth factor-inducible protein JE; RP23-350G1.3; small-inducible cytokine A2; unnamed protein product

          View more View less

          Gene Aliases: HC11; JE; MCAF; MCP-1; MCP1; Scya2; Sigje; SMC-CF

          View more View less

          UniProt ID: (Mouse) P10148

          View more View less

          Entrez Gene ID: (Mouse) 20296

          View more View less

          Function(s)
          G-protein coupled receptor binding cytokine activity protein binding chemokine activity heparin binding CCR2 chemokine receptor binding chemoattractant activity CCR chemokine receptor binding
          Process(es)
          angiogenesis response to hypoxia positive regulation of leukocyte mediated cytotoxicity positive regulation of endothelial cell proliferation leukocyte migration involved in inflammatory response chronic inflammatory response monocyte chemotaxis positive regulation of leukocyte migration positive regulation of cellular extravasation cellular calcium ion homeostasis inflammatory response immune response cytoskeleton organization transforming growth factor beta receptor signaling pathway glial cell migration regulation of cell shape response to xenobiotic stimulus response to wounding response to mechanical stimulus response to bacterium response to gamma radiation regulation of vascular endothelial growth factor production positive regulation of gene expression positive regulation of macrophage chemotaxis response to activity negative regulation of angiogenesis cytokine-mediated signaling pathway sensory perception of pain positive regulation of cell-cell adhesion positive regulation of cell migration neutrophil chemotaxis organ regeneration response to lipopolysaccharide response to progesterone positive regulation of tumor necrosis factor production cellular response to insulin stimulus positive regulation of collagen biosynthetic process response to vitamin B3 negative regulation of glial cell apoptotic process response to tumor necrosis factor helper T cell extravasation monocyte homeostasis response to isolation stress cellular response to macrophage colony-stimulating factor stimulus cellular response to platelet-derived growth factor stimulus chemokine (C-C motif) ligand 2 signaling pathway response to amino acid negative regulation of neuron apoptotic process astrocyte cell migration response to ethanol positive regulation of cell adhesion vascular endothelial growth factor receptor signaling pathway eosinophil chemotaxis macrophage chemotaxis lymphocyte chemotaxis positive regulation of synaptic transmission positive regulation of T cell activation positive chemotaxis response to glucocorticoid positive regulation of synaptic transmission, glutamatergic mammary gland involution maternal process involved in female pregnancy maternal process involved in parturition antimicrobial humoral immune response mediated by antimicrobial peptide chemokine-mediated signaling pathway cellular response to lipopolysaccharide cellular response to retinoic acid cellular response to ATP cellular response to glucose stimulus cellular response to interferon-gamma cellular response to interleukin-1 cellular response to interleukin-6 cellular response to tumor necrosis factor cellular response to estradiol stimulus cellular response to fatty acid cellular response to lipoprotein particle stimulus cellular response to high density lipoprotein particle stimulus cellular response to xenobiotic stimulus cellular response to dexamethasone stimulus positive regulation of monocyte chemotaxis positive regulation of immune complex clearance by monocytes and macrophages positive regulation of calcium ion import positive regulation of wound healing positive regulation of protein targeting to membrane energy homeostasis positive regulation of glutamate receptor signaling pathway response to cyclosporin A negative regulation of vascular endothelial cell proliferation negative regulation of G1/S transition of mitotic cell cycle positive regulation of endothelial cell apoptotic process positive regulation of apoptotic cell clearance negative regulation of natural killer cell chemotaxis
          It has to be done as per old AB suggested Products section.

          Disclaimer

          Close

          Clicking the images or links will redirect you to a website hosted by BenchSci that provides third-party scientific content. Neither the content nor the BenchSci technology and processes for selection have been evaluated by us; we are providing them as-is and without warranty of any kind, including for use or application of the Thermo Fisher Scientific products presented.

          guarantee_icon

          Performance Guarantee

          If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*

          Learn more
          help_icon

          We're here to help

          Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.

          Contact tech support
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Responsibility Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          México flag icon
          México

          Your items have has been added!


          Host server : magellan-search-blue-6b866bbc8c-vnqbn:80/100.66.76.64:80.
          git-commit: 9d76681b284ad04e8ddcd97b79462898b32e8e5b
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.61.1-2026.10.99-1.0