Search
Search
Invitrogen
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}
FIGURE: 1 / 1
This antibody detects endogenous human and mouse Annexin A13 in Western blots. In Western blots, this antibody did not cross-react with recombinant human Annexin A1, A2, A3, A4, A5, A6, A7, A8, A9, A10, or A11.
Reconstitute at 0.2 mg/mL in sterile PBS.
This gene encodes a member of the annexin family. Members of this calcium-dependent phospholipid-binding protein family play a role in the regulation of cellular growth and in signal transduction pathways. The specific function of this gene has not yet been determined; however, it is associated with the plasma membrane of undifferentiated, proliferating endothelial cells and differentiated villus enterocytes. Alternatively spliced transcript variants encoding different isoforms have been identified.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Annexin A13; Annexin XIII; annexin, intestine-specific; Annexin-13; ANXA13; Intestine-specific annexin; ISA; unnamed protein product
Gene Aliases: 1810034H17Rik; ANX13; ANXA13; AV055219; ISA
UniProt ID: (Human) P27216, (Mouse) Q99JG3
Entrez Gene ID: (Human) 312, (Mouse) 69787
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn more
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support