Search
Search
Invitrogen
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}
FIGURE: 1 / 2
Please note: We are reviewing Western blot images included in the antibody testing data in our catalog, including those provided by third parties. Unless expressly labeled or annotated as “raw-unedited”, Western blot images included in the antibody testing data in our catalog may have been edited, optimized or otherwise adjusted for presentation.
In direct ELISAs, less than 1% cross-reactivity with recombinant mouse (rm) TNF RI and rmTNF RII is observed.
Reconstitute at 0.2 mg/mL in sterile PBS.
The TNFRH3 gene encodes a protein that is part of the tumor necrosis factor receptor superfamily, which is involved in regulating immune responses, inflammation, and apoptosis. TNFRH3, also identified as tumor necrosis factor receptor homolog 3, is characterized by its similarities to other TNF receptors, yet its specific ligand and signaling pathways remain an area of active research. TNF receptors typically engage in critical interactions with ligands such as TNF-α, facilitating cellular responses that include the modulation of immune cell function, survival, and proliferation. Although TNFRH3's precise function is not fully elucidated, it is hypothesized to play a role in modulating immune signaling and inflammatory responses, potentially impacting autoimmune disease processes and cancer progression by influencing apoptosis and cell survival. Like other members of the TNF receptor family, TNFRH3 might interact with adaptor proteins to form signaling complexes that propagate downstream effects critical for immune homeostasis. Research efforts continue to define its role in physiological and pathological states, aiming to leverage its function for therapeutic applications in treating inflammatory diseases and cancer.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: mTNFRH3; TNF receptor homolog 3; TNFR; Tumor necrosis factor receptor; tumor necrosis factor receptor superfamily, member 24; unnamed protein product
Gene Aliases: Tnfrh3; Tnfrsf24
UniProt ID: (Mouse) P83626
Entrez Gene ID: (Mouse) 244237
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn more
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support