Search
Search
Invitrogen
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}
FIGURE: 1 / 22
Please note: We are reviewing Western blot images included in the antibody testing data in our catalog, including those provided by third parties. Unless expressly labeled or annotated as “raw-unedited”, Western blot images included in the antibody testing data in our catalog may have been edited, optimized or otherwise adjusted for presentation.
Expected from sequence similarity: Mouse.
In IHC-P, staining is very strong in the nucleus with some cytoplasmic staining. Prior to immunostaining paraffin tissues, antigen retrieval with sodium citrate buffer (pH 6.0) is recommended.
PGC1 alpha is a transcriptional coactivator that regulates the genes involved in energy metabolism. It interacts with PPARgamma, which permits it's interaction with multiple transcription factors. PGC-1 Alpha can interact with, and regulate the activities of, cAMP response element binding protein (CREB) and nuclear respiratory factors (NRFs). It provides a direct link between external physiological stimuli and the regulation of mitochondrial biogenesis, and is a major factor that regulates muscle fiber type determination. It may be also involved in controlling blood pressure, regulating cellular cholesterol homoeostasis, and the development of obesity.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: L-PGC-1alpha; ligand effect modulator-6; peroxisome proliferative activated receptor gamma coactivator 1; peroxisome proliferative activated receptor, gamma, coactivator 1 alpha; peroxisome proliferator activated receptor gamma coactivator 1; peroxisome proliferator activated receptor gamma coactivator-1; peroxisome proliferator activated receptor gamma, coactivator 1 alpha; peroxisome proliferator-activated receptor gamma coactivator 1 alpha; Peroxisome proliferator-activated receptor gamma coactivator 1-alpha; peroxisome proliferator-activated receptor gamma, coactivator 1 alpha; PGC-1; pgc-1 alpha; PGC-1-alpha; PGC-1a-like; PGC-1alpha; PPAR gamma coactivator 1; PPAR gamma coactivator 1alpha variant form-1; PPAR gamma coactivator variant form; PPAR gamma coactivator-1; PPAR-gamma coactivator; PPAR-gamma coactivator 1-alpha; PPARAGCIalpha; PPARgamma coactivator 1alpha; PPARGC-1-alpha; Ppargc1a-vf1; transcriptional coactivator; transcriptional coactivator; PGC-1; unnamed protein product
Gene Aliases: A830037N07Rik; Gm11133; I79_015506; LEM6; LRPGC1; PGC-1; PGC-1(alpha); PGC-1alpha; PGC-1v; PGC1; PGC1A; Pgco1; PGCvf; PGCvf-1; PGCvf1; PPARGC-1; PPARGC-1-alpha; PPARGC1
UniProt ID: (Pig) Q865B6, (Human) Q9UBK2, (Mouse) O70343, (Rat) Q9QYK2
Entrez Gene ID: (Goat) 100861288, (Pig) 397013, (Sheep) 443270, (Human) 10891, (Mouse) 19017, (Chinese hamster) 100753651, (Rat) 83516
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn more
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support