Search
Search
Invitrogen
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}
FIGURE: 1 / 9
Reconstitute with 0.2 mL of distilled water to yield a concentration of 500 µg/mL.
Positive Control - WB: rat brain tissue, rat lung tissue, mouse stomach tissue, mouse kidney tissue. ICC/IF: U20S cells. Flow: HeLa cell.
Cytosolic and membrane-bound forms of glutathione S-transferase are encoded by two distinct supergene families. At present, eight distinct classes of the soluble cytoplasmic mammalian glutathione S-transferases have been identified: alpha, kappa, mu, omega, pi, sigma, theta and zeta. This gene encodes a glutathione S-transferase that belongs to the mu class. The mu class of enzymes functions in the detoxification of electrophilic compounds, including carcinogens, therapeutic drugs, environmental toxins and products of oxidative stress, by conjugation with glutathione. The genes encoding the mu class of enzymes are organized in a gene cluster on chromosome 1p13. 3 and are known to be highly polymorphic. These genetic variations can change an individual's susceptibility to carcinogens and toxins as well as affect the toxicity and efficacy of certain drugs. Null mutations of this class mu gene have been linked with an increase in a number of cancers, likely due to an increased susceptibility to environmental toxins and carcinogens. Multiple protein isoforms are encoded by transcript variants of this gene.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: glutathione S-alkyltransferase; glutathione S-aralkyltransferase; glutathione S-aryltransferase; Glutathione S-transferase GT8.7; glutathione S-transferase M1; Glutathione S-transferase Mu 1; Glutathione S-transferase Yb-1; glutathione S-transferase Yb-1 subunit; glutathione-S-transferase mu type 1 (Yb1); glutathione-S-transferase, mu 1; glutathione-S-transferase, mu type 1 (Yb1); GST 1-1; GST 3-3; GST class-mu 1; GST HB subunit 4; GST Yb1; GSTM1; GSTM1-1; GSTM1a-1a; GSTM1b-1b; GTH4; HB subunit 4; pmGT10; S-(hydroxyalkyl)glutathione lyase; unnamed protein product
Gene Aliases: GST1; GSTA3; Gstb-1; Gstb1; GSTM1; GSTM1-1; GSTM1a-1a; GSTM1b-1b; GTH4; GTM1; H-B; MU; MU-1
UniProt ID: (Human) P09488, (Mouse) Q58ET5, (Rat) P04905
Entrez Gene ID: (Human) 2944, (Mouse) 14862, (Rat) 24423
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn more
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support