Search
Search
Invitrogen
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}
Please note: We are reviewing Western blot images included in the antibody testing data in our catalog, including those provided by third parties. Unless expressly labeled or annotated as “raw-unedited”, Western blot images included in the antibody testing data in our catalog may have been edited, optimized or otherwise adjusted for presentation.
Reconstitute with 0.2 mL of distilled water to yield a concentration of 500 µg/mL.
Positive Control - WB: human Hela whole cell, human HepG2 whole cell. IHC: mouse intestine tissue, rat intestine tissue, human lung cancer tissue.
This gene encodes a receptor for inositol 1,4,5-trisphosphate, a second messenger that mediates the release of intracellular calcium. The receptor contains a calcium channel at the C-terminus and the ligand-binding site at the N-terminus. Knockout studies in mice suggest that type 2 and type 3 inositol 1,4,5-trisphosphate receptors play a key role in exocrine secretion underlying energy metabolism and growth.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: inositol 1,4,5-triphosphate receptor 3; inositol 1,4,5-triphosphate receptor, type 3; inositol 1,4,5-trisphosphate receptor type 3; insP3R3; ion channel; IP3 receptor; IP3R 3; type 3 inositol 1,4,5-trisphosphate receptor; type 3 InsP3 receptor; unnamed protein product
Gene Aliases: CMT1J; IMD132; IMD133; IP3R; IP3R 3; IP3R-3; IP3R3; IP3R3X; Itpr-3; tf
UniProt ID: (Human) Q14573, (Mouse) P70227, (Rat) Q63269
Entrez Gene ID: (Human) 3710, (Mouse) 16440, (Rat) 25679
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn more
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support