Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          • Primary Antibodies ›
          • Laminin gamma-1,2,3 Antibodies

          Invitrogen

          Laminin gamma-1,2,3 Polyclonal Antibody

          Advanced Verification
          View all (2) Laminin gamma-1,2,3 antibodies

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}

          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Tech Support
          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Tech Support
          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Tech Support

          Cite Laminin gamma-1,2,3 Polyclonal Antibody

          Additional Information:
          {{banner.description}}
          {{$ctrl.isSkuNotesCollapsed ? 'View more' : 'View less'}}
          • Antibody Testing Data (5)
          • Advanced Verification (2)
          Laminin gamma-1,2,3 Antibody in Immunocytochemistry (ICC/IF)
          Group 53 Created with Sketch.
          Laminin gamma-1,2,3 Antibody in Immunocytochemistry (ICC/IF)
          Group 53 Created with Sketch.

          FIGURE: 1 / 7

          {{ $ctrl.currentElement.advancedVerification.fullName }}
          {{ $ctrl.currentElement.advancedVerification.text }}

          Laminin gamma-1,2,3 Antibody (PA5-79579) in ICC/IF

          Immunofluorescence analysis of Laminin gamma-1,2,3 was performed using 70% confluent log phase Hep G2 cells. The cells were fixed with 4% paraformaldehyde for 10 minutes, permeabilized with 0.1% Triton™ X-100 for 15 minutes and blocked with 2% BSA for 1 hour at room temperature. The cells were labeled with Laminin gamma-1,2,3 Polyclonal Antibody (Product # PA5-79580) at 1:100 dilution in 0.1% BSA, incubated... View More {{ $ctrl.currentElement.advancedVerification.fullName }} validation info. View more
          Published figure supplied by benchsci-logo
          PMID: {{$ctrl.currentElement.benchSciPubmedId}}
          View Product

          {{$ctrl.videoDesc}}

          {{$ctrl.videoLongDesc}}

          Laminin gamma-1,2,3 Antibody in Immunocytochemistry (ICC/IF)
          Laminin gamma-1,2,3 Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          Laminin gamma-1,2,3 Antibody in Immunohistochemistry (Frozen) (IHC (F))
          Laminin gamma-1,2,3 Antibody in Western Blot (WB)
          Laminin gamma-1,2,3 Antibody in Western Blot (WB)
          Laminin gamma-1,2,3 Antibody
          Laminin gamma-1,2,3 Antibody
          Laminin gamma-1,2,3 Polyclonal Antibody

          Product Details

          PA5-79579

          Applications
          Tested Dilution
          Publications

          Western Blot (WB)

          0.1-0.5 µg/mL
          -

          Immunohistochemistry (Paraffin) (IHC (P))

          0.5-1 µg/mL
          -

          Immunohistochemistry (Frozen) (IHC (F))

          0.5-1 µg/mL
          -

          Immunocytochemistry (ICC/IF)

          0.5-1 µg/mL
          -
          Product Specifications

          Species Reactivity

          Human, Mouse, Rat

          Host/Isotype

          Rabbit / IgG

          Class

          Polyclonal

          Type

          Antibody

          Immunogen

          Peptide mixture of laminin gamma1,2,3 (DLNKLNEIEGTLNKAKDEMK; MSELEERARQQRGHLHLLET; LQRKLSLLEQESQQQELQIQ).
          3D Epitope / Immunogen

          Conjugate

          Unconjugated Unconjugated Unconjugated

          Form

          Lyophilized

          Concentration

          500 µg/mL

          Amount

          100 µg

          Purification

          Antigen affinity chromatography

          Storage buffer

          5mg BSA, 0.9mg NaCl, 0.2mg Na2PO4

          Contains

          0.05mg sodium azide

          Storage conditions

          -20°C

          Shipping conditions

          Ambient (domestic); Wet ice (international)

          RRID

          AB_2746694

          Product Specific Information

          This antibody reacts will all 3 isoforms of Laminin gamma: Laminin gamma-1, gamma-2 and gamma-3.

          Reconstitute with 0.2 mL of distilled water to yield a concentration of 500 µg/mL.

          Positive Control - WB: human A431 whole cell, human A549 whole cell, human PC-3 whole cell, human Caco-2 whole cellrat NRK whole cell, rat RH35 whole cell, mouse NIH/3T3 whole cell, mouse Hepa1-6 whole cell. IHC: human lung cancer tissue IHC-F: human placenta tissue.

          Target Information

          Extracellular matrix (ECM) surrounds and separates most tissue compartments, determines tissue architecture and dynamically interacts with its surrounding cells. The basement membrane is a part of the ECM and is composed of collagens, proteoglycans, elastin and several glycoproteins of which laminin is the most abundant. Laminins are large heterotrimeric, noncollagenous glycoproteins composed of alpha, beta, and gamma chains Dystroglycan is a major laminin receptor in muscle and epithelia, and nidogen is a high-affinity laminin-binding protein important for basement-membrane assembly.

          For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.

          References (0)

          Have you cited this product in a publication?
          Let us know so we can reference it here.
          Cite this product

          Bioinformatics

          Protein Aliases: basement membrane protein; BM600-100kDa; Cell-scattering factor 140 kDa subunit; CSF 140 kDa subunit; Epiligrin subunit gamma; Kalinin subunit gamma; Kalinin/nicein/epiligrin 100 kDa subunit; Ladsin 140 kDa subunit; lamimin, gamma 2; Laminin B2 chain; laminin B2 precursor; Laminin B2t chain; laminin gamma2 chain; Laminin subunit gamma-1; Laminin subunit gamma-2; Laminin subunit gamma-3; laminin, gamma 1 (formerly LAMB2); laminin, gamma 2; laminin, gamma 3; Laminin-1 subunit gamma; Laminin-10 subunit gamma; Laminin-11 subunit gamma; Laminin-12 subunit gamma; Laminin-14 subunit gamma; Laminin-15 subunit gamma; Laminin-2 subunit gamma; Laminin-3 subunit gamma; Laminin-4 subunit gamma; Laminin-5 subunit gamma; Laminin-6 subunit gamma; Laminin-7 subunit gamma; Laminin-8 subunit gamma; Laminin-9 subunit gamma; Large adhesive scatter factor 140 kDa subunit; Nicein subunit gamma; nicein, 100kD; nicein, 100kDa; S-LAM gamma; S-laminin subunit gamma; unnamed protein product

          View more View less

          Gene Aliases: AA589349; AI562206; AW240805; B2e; B2T; BM600; CSF; EBR2; EBR2A; JEB3A; JEB3B; LAMB2; LAMB2T; LAMC1; LAMC2; LAMC3; LAMNB2; OCCM

          View more View less

          UniProt ID: (Human) P11047, (Human) Q13753, (Human) Q9Y6N6, (Mouse) Q9R0B6

          View more View less

          Entrez Gene ID: (Human) 3915, (Human) 3918, (Human) 10319, (Mouse) 226519, (Rat) 117036, (Mouse) 16782, (Rat) 192362, (Rat) 311862, (Mouse) 23928

          View more View less

          Function(s)
          extracellular matrix structural constituent extracellular matrix constituent conferring elasticity heparin binding structural molecule activity protein binding glycosphingolipid binding molecular_function extracellular matrix protein DNA metabolism protein
          Process(es)
          cell adhesion endoderm development tissue development cell migration extracellular matrix disassembly hemidesmosome assembly substrate adhesion-dependent cell spreading maintenance of blood-brain barrier positive regulation of cell adhesion positive regulation of epithelial cell proliferation positive regulation of muscle cell differentiation protein-containing complex assembly regulation of basement membrane organization positive regulation of integrin-mediated signaling pathway positive regulation of cell proliferation epidermis development positive regulation of cell migration cell morphogenesis visual perception astrocyte development retina development in camera-type eye protein complex assembly neuron projection development biological_process cell morphogenesis involved in differentiation
          It has to be done as per old AB suggested Products section.

          Disclaimer

          Close

          Clicking the images or links will redirect you to a website hosted by BenchSci that provides third-party scientific content. Neither the content nor the BenchSci technology and processes for selection have been evaluated by us; we are providing them as-is and without warranty of any kind, including for use or application of the Thermo Fisher Scientific products presented.

          guarantee_icon

          Performance Guarantee

          If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*

          Learn more
          help_icon

          We're here to help

          Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.

          Contact tech support
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Responsibility Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          México flag icon
          México

          Your items have has been added!


          Host server : magellan-search-green-794c4b9cd6-gkxk9:80/100.66.77.77:80.
          git-commit: 7e8dd63fde7d4486b98ef9ac5cf4e365aa842ffe
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.52.0-Offline