Search
Search
Invitrogen
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}
Please note: We are reviewing Western blot images included in the antibody testing data in our catalog, including those provided by third parties. Unless expressly labeled or annotated as “raw-unedited”, Western blot images included in the antibody testing data in our catalog may have been edited, optimized or otherwise adjusted for presentation.
Purity is > 95% by SDS-PAGE.
PSA is a chymotrypsin-like serine protease (kallikrein family) produced by the prostate epithelium, and is abundant in seminal fluid. PSA can be detected in the sera of patients with prostatic carcinoma. It is predominantly complexed to a liver-derived serine protease inhibitor, alpha-1-antichymotrypsin (ACT). A higher proportion of serum PSA is complexed to ACT in prostate cancer than in benign prostate hyperplasia. PSA is used to confirm prostatic acinar cell origin in primary and metastatic carcinoma and to rule out non-prostatic carcinoma mimics.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: 7S nerve growth factor gamma chain; 7S nerve growth factor gamma subunit; Antigen; gamma-NGF; gamma-seminoprotein; glandular kallikrein K3; kallikrein 1; Kallikrein 3; kallikrein a4; kallikrein-3; Kallikrein-3 (KLK3); KLK-3; nerve growth factor gamma chain; nerve growth factor gamma polypeptide; P-30 antigen; pancreatic kallikrein; prostate specific antigen precursor; prostate-specific (APS); prostate-specific antigen; PS kallikrein; PSA; KLK3; semenogelase; seminin; tissue kallikrein-3; true tissue kallikrein; unnamed protein product
Gene Aliases: APS; gamma-NGF; hK3; Klk1; Klk1c1; KLK2A1; Klna4; mGk-3; Ngfg; PSA; rGK-1; rK-1
UniProt ID: (Human) P07288, (Mouse) P00756, (Rat) P00758
Entrez Gene ID: (Human) 354, (Mouse) 18050, (Rat) 24594
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn more
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support