Search
Search
Invitrogen
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}
FIGURE: 1 / 1
Please note: We are reviewing Western blot images included in the antibody testing data in our catalog, including those provided by third parties. Unless expressly labeled or annotated as “raw-unedited”, Western blot images included in the antibody testing data in our catalog may have been edited, optimized or otherwise adjusted for presentation.
Immunogen sequence: MAEPSSARRP VPLIESELYF LIARYLSAGP CRRAAQVLVQ ELEQYQLLPK RLDWEGNEHN RSYEELVLSN KHVAPDHLLQ ICQRIGPMLD KEIPPSISRV TSLLGAGRQS LLRTAKGTLI; Positive Samples: HeLa, Jurkat, SGC-7901, NIH/3T3; Cellular Location: Cytoplasm, Nucleus
WDR9.3 encodes a member of the WD repeat protein family. WD repeats are minimally conserved regions of approximately 40 amino acids typically bracketed by gly-his and trp-asp (GH-WD), which may facilitate formation of heterotrimeric or multiprotein complexes. Members of this family are involved in a variety of cellular processes, including cell cycle progression, signal transduction, apoptosis, and gene regulation. This protein contains 2 bromodomains and multiple WD repeats, and the function of this protein is not known. This gene is located within the Down syndrome region 2 on chromosome 21. Alternative splicing of this gene generates 3 transcript variants diverging at the 3' ends.
⚠WARNING: This product can expose you to chemicals including mercury, which is known to the State of California to cause birth defects or other reproductive harm. For more information go to www.P65Warnings.ca.gov.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: FLJ43918; transcriptional unit N143; unnamed protein product; WD repeat domain 9; WD repeat protein WDR9-form2; WD repeat-containing protein 9
Gene Aliases: 5330419I02Rik; C21orf107; CILD51; D530019K20Rik; DCAF19; G1-403-16; N143; repro5; WDR9; WRD9
UniProt ID: (Human) Q9NSI6, (Mouse) Q921C3
Entrez Gene ID: (Human) 54014, (Mouse) 93871
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn more
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support