Search
Search
Invitrogen
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}
FIGURE: 1 / 1
Please note: We are reviewing Western blot images included in the antibody testing data in our catalog, including those provided by third parties. Unless expressly labeled or annotated as “raw-unedited”, Western blot images included in the antibody testing data in our catalog may have been edited, optimized or otherwise adjusted for presentation.
Immunogen sequence: DKGDPHLCDF LETYYLNEQV KSIKELGDHV HNLVKMGAPD AGLAEYLFDT HTLGNENKQN; Positive Samples: Mouse testis; Cellular Location: Mitochondrion
FTMT stores iron in a soluble, non-toxic, readily available form. FTMT is important for iron homeostasis and has ferroxidase activity. Iron is taken up in the ferrous form and deposited as ferric hydroxides after oxidation.
⚠WARNING: This product can expose you to chemicals including mercury, which is known to the State of California to cause birth defects or other reproductive harm. For more information go to www.P65Warnings.ca.gov.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: ferritin H; ferritin heavy chain 3; unnamed protein product
Gene Aliases: 4930447C24Rik; FerH; Fth3; MtF
UniProt ID: (Mouse) Q9D5H4
Entrez Gene ID: (Mouse) 67634
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn more
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support