Search
Search
Invitrogen
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}
Please note: We are reviewing Western blot images included in the antibody testing data in our catalog, including those provided by third parties. Unless expressly labeled or annotated as “raw-unedited”, Western blot images included in the antibody testing data in our catalog may have been edited, optimized or otherwise adjusted for presentation.
Immunogen sequence: GILNLNKMVK QVTGKMPILS YWPYGCHCGL GGRGQPKDAT DWCCQTHDCC YDHLKTQGCS IYKDYYRYNF SQGNIHCSDK GSWCEQQLCA CDKEVAFCLK RNLDTYQKRL RFYWRPHCRG QTPGC; Positive Samples: HL-60, THP-1
PA2 catalyzes the calcium-dependent hydrolysis of the 2- acyl groups in 3-sn-phosphoglycerides. L-alpha-1-palmitoyl-2- linoleoyl phosphatidylethanolamine is more efficiently hydrolyzed than the other phospholipids examined.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: GIID sPLA2; group IID sPLA2; phosphatidylcholine 2-acylhydrolase 2D; PLA2IID; secretory phospholipase A2s; secretory-type PLA, stroma-associated homolog; SPLASH; unnamed protein product
Gene Aliases: GIID; GIID sPLA2; mGIID; PLA2IID; sPLA2-IID; sPLA2S; SPLASH
UniProt ID: (Human) Q9UNK4, (Mouse) Q9WVF6
Entrez Gene ID: (Human) 26279, (Mouse) 18782, (Rat) 298579
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn more
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support