Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • New Products
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          • Primary Antibodies ›
          • LAP2 Antibodies

          Invitrogen

          LAP2 Polyclonal Antibody

          1 Published Figure
          1 Reference
          View all (11) LAP2 antibodies

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}

          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Datasheet
          Certificates
          Protocols
          Questions & Answers

          Cite LAP2 Polyclonal Antibody

          {{$ctrl.translations['antibody.pdp.skuNotes.additionalInformation']}}:

          {{$ctrl.translations[$ctrl.isSkuNotesCollapsed ? 'antibody.pdp.skuNotes.viewMore' : 'antibody.pdp.skuNotes.viewLess']}}
          • Antibody Testing Data (7)
          • Published Figures (1)
          LAP2 Antibody in Immunocytochemistry (ICC/IF)
          Group 53 Created with Sketch.
          LAP2 Antibody in Immunocytochemistry (ICC/IF)
          Group 53 Created with Sketch.

          FIGURE: 1 / 8

          {{ $ctrl.currentElement.advancedVerification.fullName }}
          {{ $ctrl.currentElement.advancedVerification.text }}

          LAP2 Antibody (PA5-96154) in ICC/IF

          Immunofluorescence analysis of LAP2 in NIH/3T3 cells. Samples were incubated with LAP2 Polyclonal antibody (Product # PA5-96154) using a dilution of 1:50 (40x lens). Blue: DAPI for nuclear staining. {{ $ctrl.currentElement.advancedVerification.fullName }} validation info. View more
          Published figure supplied by benchsci-logo
          PMID: {{$ctrl.currentElement.benchSciPubmedId}}
          View Product

          {{$ctrl.videoDesc}}

          {{$ctrl.videoLongDesc}}

          LAP2 Antibody in Immunocytochemistry (ICC/IF)
          LAP2 Antibody in Immunocytochemistry (ICC/IF)
          LAP2 Antibody in Immunocytochemistry (ICC/IF)
          LAP2 Antibody in Immunocytochemistry (ICC/IF)
          LAP2 Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          LAP2 Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          LAP2 Antibody in Western Blot (WB)
          LAP2 Antibody in Western Blot (WB)

          Please note: We are reviewing Western blot images included in the antibody testing data in our catalog, including those provided by third parties. Unless expressly labeled or annotated as “raw-unedited”, Western blot images included in the antibody testing data in our catalog may have been edited, optimized or otherwise adjusted for presentation.

          LAP2 Polyclonal Antibody

          Product Details

          PA5-96154

          Applications
          Tested Dilution
          Publications

          Western Blot (WB)

          1:500-1:1,000
          View 1 publication 1 publication

          Immunohistochemistry (Paraffin) (IHC (P))

          1:50-1:200
          -

          Immunocytochemistry (ICC/IF)

          1:50-1:200
          -

          ELISA (ELISA)

          1 µg/mL
          -
          Product Specifications

          Species Reactivity

          Human, Mouse, Rat

          Published species

          Human

          Host/Isotype

          Rabbit / IgG

          Class

          Polyclonal

          Type

          Antibody

          Immunogen

          Recombinant protein (or fragment)
          3D Epitope / Immunogen

          Conjugate

          Unconjugated Unconjugated Unconjugated

          Form

          Liquid

          Concentration

          0.21 mg/mL

          Amount

          21 µg

          Purification

          Affinity Chromatography

          Storage buffer

          PBS, pH 7.3, with 50% glycerol

          Contains

          0.05% ProClin 300

          Storage conditions

          -20°C, Avoid Freeze/Thaw Cycles

          Shipping conditions

          Ambient (domestic); Wet ice (international)

          RRID

          AB_2807956

          Product Specific Information

          Immunogen sequence: MPEFLEDPSV LTKDKLKSEL VANNVTLPAG EQRKDVYVQL YLQHLTARNR PPLPAGTNSK GPPDFSSDEE REPTPVLGSG AAAAGRSRAA VGRKATKKTD KPRQEDKDDL DVTELTNEDL LDQLVKYGVN PGPIVGTTRK LYEKKLLKLR EQGTESRSST PLPTISSSAE NTRQNGSNDS DRYSDNEEDS KIELKLEKRE PLKGRAKTPV TLKQRRVEHN QSYSQAGITE TEWTSGSSKG GPLQALTRES TRGSRRTPRK RVETSEHFRI DGPVISESTP IAETIMASSN ESLVVNRVTG NFKHASPILP ITEFSDIPRR APKKPLTRAE VGEKTEERRV ERDILKEMFP YEASTPTGIS ASCRRPIKGA AGRPLELSDF RMEESFSSKY VPKYVPLADV KSEKTKKGRS; Positive Samples: 22Rv1, K-562, HepG2, 293T, SH-SY5Y, H460; Cellular Location: Chromosome, Nucleus

          Target Information

          In eukaryotic cells, the nuclear envelope consists of inner and outer membranes which define the border between the nucleus and cytoplasm. The inner and outer nuclear membranes are joined by large, supramolecular assemblies called nuclear pore complexes through which various molecules pass into and out of the nucleus. The inner membrane is lined by a filamentous meshwork called the nuclear lamina made up of proteins called lamins. The attachment of the lamins to the inner nuclear membrane appears to occur through their association with 4 related integral membrane proteins called Lamin Associated Polypeptides (LAPs). LAP 1A, 1B and 1C share a high degree of sequence homology while LAP 2 is distinct from the others. LAP 1A/B bind lamins A, C and B1, while LAP 2 binds lamin B1 only. In addition to lamin binding, LAP 2 has been shown to bind to chromosomes. Phosphorylation of LAP 2 during mitosis inhibits binding to both lamins and chromosomes suggesting that LAP 2 plays an important role in nuclear envelope re-assembly.

          For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.

          Bioinformatics

          Protein Aliases: lamina-associated polypeptide 2; LEM domain containing 4; similar to human thymopoietin gamma encoded by GenBank Accession Number U09088; Thymopoietin (lamina associated polypeptide 2); thymopoietin-related peptide; TP beta; unnamed protein product

          View more View less

          Gene Aliases: 5630400D24Rik; CMD1T; LAP2; LEMD4; PRO0868; TP

          View more View less

          UniProt ID: (Human) P42166, (Mouse) Q61029, (Rat) Q62733

          View more View less

          Entrez Gene ID: (Human) 7112, (Mouse) 21917, (Rat) 25359

          View more View less

          Function(s)
          DNA binding protein binding lamin binding cadherin binding identical protein binding
          Process(es)
          regulation of transcription, DNA-templated nuclear envelope reassembly
          It has to be done as per old AB suggested Products section.

          Disclaimer

          Close

          Clicking the images or links will redirect you to a website hosted by BenchSci that provides third-party scientific content. Neither the content nor the BenchSci technology and processes for selection have been evaluated by us; we are providing them as-is and without warranty of any kind, including for use or application of the Thermo Fisher Scientific products presented.

          guarantee_icon

          Performance Guarantee

          If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*

          Learn more
          help_icon

          We're here to help

          Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.

          Contact tech support
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-green-5db85fbf84-7kv6q:80/100.66.72.150:80.
          git-commit: 64d6f4084a597c0a860dbc264f43a6f389f0d7ac
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.60.1-2026-secondary-format-patch