Search
Search
Invitrogen
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}
FIGURE: 1 / 1
Immunogen sequence: DDGGASGRPL PKAQPGHRSY NLQERRRIGS MTGAEQALLP RVPTDEIEAQ TLATADLDLM KSHRFEDVPG VRRHLVRKNA KGSTQSGREG REPGPTPRAR P; Positive Samples: HeLa, A-549, Jurkat, HT-29; Cellular Location: Membrane, Multi-pass membrane protein
Plasma membrane anion exchange protein of wide distribution.
⚠WARNING: This product can expose you to chemicals including mercury, which is known to the State of California to cause birth defects or other reproductive harm. For more information go to www.P65Warnings.ca.gov.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: AE 2; Anion exchange protein 2; anion exchanger 2 type a; anion exchanger 2 type b1; anion exchanger 2 type b2; BND3L; Erythrocyte band 3; erythrocyte membrane protein band 3-like 1; Non-erythroid band 3-like protein; solute carrier family 4 (anion exchanger), member 2; Solute carrier family 4 member 2; unnamed protein product
Gene Aliases: AE2; BND3L; EPB3L1; HKB3; MPB3L; NBND3; OPTB9; SLC4A2
UniProt ID: (Human) P04920
Entrez Gene ID: (Human) 6522
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn more
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support