Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          • Primary Antibodies ›
          • Synaptotagmin 4 Antibodies

          Invitrogen

          Synaptotagmin 4 Polyclonal Antibody

          View all (25) Synaptotagmin 4 antibodies

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}

          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Tech Support
          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Tech Support
          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Tech Support

          Cite Synaptotagmin 4 Polyclonal Antibody

          Additional Information:
          {{banner.description}}
          {{$ctrl.isSkuNotesCollapsed ? 'View more' : 'View less'}}
          • Antibody Testing Data (4)
          Synaptotagmin 4 Antibody in Immunocytochemistry (ICC/IF)
          Group 53 Created with Sketch.
          Synaptotagmin 4 Antibody in Immunocytochemistry (ICC/IF)
          Group 53 Created with Sketch.

          FIGURE: 1 / 4

          {{ $ctrl.currentElement.advancedVerification.fullName }}
          {{ $ctrl.currentElement.advancedVerification.text }}

          Synaptotagmin 4 Antibody (PA5-96781) in ICC/IF

          Immunofluorescence analysis of Synaptotagmin 4 in U2OS cells. Samples were incubated with Synaptotagmin 4 Polyclonal antibody (Product # PA5-96781) using a dilution of 1:100. Blue: DAPI for nuclear staining. {{ $ctrl.currentElement.advancedVerification.fullName }} validation info. View more
          Published figure supplied by benchsci-logo
          PMID: {{$ctrl.currentElement.benchSciPubmedId}}
          View Product

          {{$ctrl.videoDesc}}

          {{$ctrl.videoLongDesc}}

          Synaptotagmin 4 Antibody in Immunocytochemistry (ICC/IF)
          Synaptotagmin 4 Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          Synaptotagmin 4 Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          Synaptotagmin 4 Antibody in Western Blot (WB)
          Synaptotagmin 4 Polyclonal Antibody

          Product Details

          PA5-96781

          Applications
          Tested Dilution
          Publications

          Western Blot (WB)

          1:500-1:1,000
          -

          Immunohistochemistry (Paraffin) (IHC (P))

          1:50-1:200
          -

          Immunocytochemistry (ICC/IF)

          1:50-1:200
          -

          ELISA (ELISA)

          1 µg/mL
          -
          Product Specifications

          Species Reactivity

          Human, Mouse, Rat

          Host/Isotype

          Rabbit / IgG

          Class

          Polyclonal

          Type

          Antibody

          Immunogen

          Recombinant fusion protein containing a sequence corresponding to amino acids 38-190 of human SYT4 (NP_0658341)
          3D Epitope / Immunogen

          Conjugate

          Unconjugated Unconjugated Unconjugated

          Form

          Liquid

          Concentration

          0.71 mg/mL

          Amount

          71 µg

          Purification

          Affinity Chromatography

          Storage buffer

          PBS, pH 7.3, with 50% glycerol

          Contains

          0.02% sodium azide

          Storage conditions

          -20°C, Avoid Freeze/Thaw Cycles

          Shipping conditions

          Ambient (domestic); Wet ice (international)

          RRID

          AB_2808583

          Product Specific Information

          Immunogen sequence: CQRKSSKSNK TPPYKFVHVL KGVDIYPENL NSKKKFGADD KNEVKNKPAV PKNSLHLDLE KRDLNGNFPK TNLKPGSPSD LENATPKLFL EGEKESVSPE SLKSSTSLTS EEKQEKLGTL FFSLEYNFER KAFVVNIKEA RGLPAMDEQS MTS; Positive Samples: SH-SY5Y, NIH/3T3, Mouse brain, Mouse spinal cord, Mouse eye, Rat brain; Cellular Location: Cytoplasmic vesicle, Single-pass membrane protein, secretory vesicle, synaptic vesicle membrane

          Target Information

          May be involved in Ca2+-dependent exocytosis of secretory vesicles through Ca2+ and phospholipid binding to the C2 domain or may serve as Ca2+ sensors in the process of vesicular trafficking and exocytosis. Plays a role in dendrite formation by melanocytes (PubMed:23999003).

          For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.

          References (0)

          Have you cited this product in a publication?
          Let us know so we can reference it here.
          Cite this product

          Bioinformatics

          Protein Aliases: synaptotagmin 4; Synaptotagmin IV; Synaptotagmin-4; Synaptotagmin4; SytIV; unnamed protein product

          View more View less

          Gene Aliases: HsT1192; KIAA1342; Syt3; SYT4; SytIV

          View more View less

          UniProt ID: (Human) Q9H2B2, (Rat) P50232, (Mouse) P40749

          View more View less

          Entrez Gene ID: (Human) 6860, (Rat) 64440, (Mouse) 20983

          View more View less

          Function(s)
          SNARE binding calcium ion binding protein binding calcium-dependent phospholipid binding metal ion binding calcium ion sensor activity phosphatidylserine binding transporter activity syntaxin binding clathrin binding membrane trafficking regulatory protein
          Process(es)
          vesicle fusion neurotransmitter secretion memory positive regulation of glutamate secretion regulation of dopamine secretion vesicle-mediated transport regulation of calcium ion-dependent exocytosis regulation of endocytosis cell differentiation regulation of vesicle fusion negative regulation of catecholamine secretion negative regulation of calcium ion-dependent exocytosis positive regulation of calcium ion-dependent exocytosis negative regulation of short-term neuronal synaptic plasticity negative regulation of protein secretion calcium-dependent activation of synaptic vesicle fusion dense core granule cytoskeletal transport regulation of trans-synaptic signaling by BDNF, modulating synaptic transmission regulation of postsynaptic dense core vesicle exocytosis negative regulation of secretion by cell positive regulation of dendrite extension positive regulation of dense core granule exocytosis negative regulation of retrograde trans-synaptic signaling by neuropeptide negative regulation of synaptic vesicle exocytosis calcium ion regulated exocytosis negative regulation of vesicle fusion exocytosis synaptic vesicle exocytosis synaptic vesicle transport calcium ion-regulated exocytosis of neurotransmitter
          It has to be done as per old AB suggested Products section.

          Disclaimer

          Close

          Clicking the images or links will redirect you to a website hosted by BenchSci that provides third-party scientific content. Neither the content nor the BenchSci technology and processes for selection have been evaluated by us; we are providing them as-is and without warranty of any kind, including for use or application of the Thermo Fisher Scientific products presented.

          guarantee_icon

          Performance Guarantee

          If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*

          Learn more
          help_icon

          We're here to help

          Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.

          Contact tech support
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          • Price & Freight Policy
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • German Supply Chain Due Diligence Act
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Legal Notice
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Germany flag icon
          Germany

          Your items have has been added!


          Host server : magellan-search-green-794c4b9cd6-gkxk9:80/100.66.77.77:80.
          git-commit: 7e8dd63fde7d4486b98ef9ac5cf4e365aa842ffe
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.52.0-Offline