Can an existing Thermo Scientific PSM-400MPX Particle Size Monitor be upgraded to the Thermo Scientific Particle Size Analyzer PSM-500?
Yes. The Thermo Scientific PSM-500 Particle Size Analyzer is designed as an upgrade option for the discontinued Thermo Scientific PSM-400MPX Particle Size Monitor. Compared with the PSM-400MPX Particle Size Monitor, the PSM-500 Particle Size Analyzer includes updated software, a contemporary touch-panel interface, field-replaceable transducers with updated bandwidth, and updated alarm and reporting capabilities.
What features of the Thermo Scientific Particle Size Analyzer PSM-500 support maintenance and servicing?
The Thermo Scientific Particle Size Analyzer PSM-500 includes field-replaceable ultrasonic transducers to support servicing. The system typically operates without sample pumping or dilution and does not require a UPS, reducing the amount of supporting equipment required for operation.
Is the Thermo Scientific Particle Size Analyzer PSM-500 suitable for harsh mineral processing environments?
Yes. The Thermo Scientific Particle Size Analyzer PSM-500 is designed for demanding mineral processing environments. It features an IP65-rated enclosure, NEMA 4X corrosion-resistant internals, temperature compensation, and an ambient operating temperature range of -10 degrees C to +50 degrees C. Wetted components are made from engineering polymers or rubber-covered steel, and the analyzer can handle caustic slurries up to pH 12.5. Extra protection is required for use in outdoor operation or direct exposure to sunlight and ultraviolet radiation.
Can the Thermo Scientific Particle Size Analyzer PSM-500 monitor multiple process streams?
Yes. The Thermo Scientific Particle Size Analyzer PSM-500 can be optioned with the ability to analyze one, two or three process streams. This allows operators to obtain particle size information from multiple sampling points in the same plant area using a single analyzer.
Does the Thermo Scientific Particle Size Analyzer PSM-500 measure percent solids as well as particle size?
Yes. Percent solids measurement is a standard capability of the Thermo Scientific Particle Size Analyzer PSM-500. The specified sample range is 10% to 60% solids by weight and 3% to 26% solids by volume. PSM-500 size fraction models typically use percent solids for compensation. Measuring particle size distribution and percent solids gives operators additional information for monitoring mineral processing and grinding circuit conditions.
How can online particle size analysis improve grinding circuit performance?
The Thermo Scientific Particle Size Analyzer PSM-500 gives operators frequent information about changes in particle size distribution and percent solids. Operators can use these measurements when evaluating and adjusting grinding circuit conditions to maintain the desired grind size. The analyzer typically updates measurements every 10 seconds to support process monitoring and control.
How accurate is the Thermo Scientific Particle Size Analyzer PSM-500?
Data generated by the Thermo Scientific Particle Size Analyzer PSM-500 are typically accurate to better than 1% absolute. The analyzer also offers frequent, real-time measurements of particle size distribution and percent solids that operators can use when monitoring grinding circuit conditions.
What particle size range can the Thermo Scientific Particle Size Analyzer PSM-500 measure?
The Thermo Scientific Particle Size Analyzer PSM-500 has a particle size measurement range of approximately 1 to 1,000 microns, which is optimized according to the application. It can measure up to five particle size fractions simultaneously plus percent solids and typically updates measurements every 10 seconds, giving operators frequent particle size assay information for process monitoring.
How does the Thermo Scientific Particle Size Analyzer PSM-500 measure particle size?
The Thermo Scientific Particle Size Analyzer PSM-500 uses ultrasonic measurement technology to determine particle size in a process slurry. The analyzer draws a continuous sample using a vacuum eductor, de-aerates the sample in a conditioner, and passes it between two pairs of diametrically opposed ultrasonic sensors for measurement before returning it to the process. The system typically does not require sample dilution or pumping.
When should I use the Thermo Scientific Particle Size Analyzer PSM-500 for online particle size monitoring?
The Thermo Scientific Particle Size Analyzer PSM-500 is designed for real-time, in-stream particle size monitoring in mineral processing applications. It can analyze up to three process streams and measure up to five particle size fractions simultaneously plus percent solids. These measurements give operators timely information for monitoring grinding circuit conditions, mineral recovery, product quality, and energy use.