Qubit™ ssDNA Assay Kit - FAQs
Qubit™ ssDNA Assay Kit - FAQs
View additional product information for Qubit™ ssDNA Assay Kit - FAQs (Q10212)
15 product FAQs found
I'm seeing other kit-related problems besides the "Standards incorrect" message with my Qubit assay. What do you suggest I try?
Here are several suggestions:
1.View the raw fluorescence value (RFU) for the standards under “Check Standards” or “Check Calibration”. Confirm that the values for the samples fall between the values of the standards (or a little above the highest standard). If they do not, the sample is out of the accurate range of the assay. Refer to the confidence ranges for each assay in the product manuals. The readout in the assay will be to 2 significant figures instead of 3 if the assay sample is out of the high confidence range.
To bring the sample into the accurate range, dilute the sample or use more or less of it (for example, 10 µL instead of 2 µL if the sample reads low).
2.Check for temperature issues: The assay is temperature sensitive and the fluorescent signal can decrease at higher temperatures. Temperature fluctuations between samples, or between samples and standards, can cause problems. Make sure that the buffer and Qubit reagent in DMSO are at room temperature. The buffer and Qubit reagent should be stored at room temperature, not in the refrigerator. Even after 2-3 hours at room temperature, buffer previously stored at 4°C can remain below room temperature. Make sure your samples and working solution are not too warm (including those straight from a centrifuge). Samples kept in the Qubit instrument too long or read multiple times can warm up. If you want to perform multiple readings of a single tube, you should remove the tube from the instrument and let it equilibrate to room temperature for 30 seconds before taking another reading. Also, do not hold tubes in your hand for very long before reading them in the instrument, since this can warm the sample, resulting in a low reading.
3.Ensure that you have prepared the Qubit working solution correctly (1:200 dilution using the buffer provided in the kit). Ensure that you have prepared the standard tubes correctly (10 µL of each standard in 190 µL of the working solution). Ensure that the tubes are filled with at least 200 µL (both standards and samples).
4.Ensure that the reagents and standards you are using are less than 6 months old, and that the standards have been stored correctly. The Qubit reagent stock solution should be protected from light as much as possible.
5.Ensure that you have selected the correct assay on the Qubit Fluorometer for the Qubit assay you are performing.
6.Ensure that the lid is completely closed when reading standards and samples.
7.Use recommended tubes (both so the tube does not obstruct the lid, and for optical clarity). Some types of tubes can have high autofluorescence that will affect the assay.
8.Did you enter the number of microliters of stock you pipetted into the working solution into the Qubit instrument? If so, the reading after giving the Qubit Fluorometer this information is the concentration of your stock solution. If you did not, the reading you got is for the concentration in the assay tube (the tube you put into the Qubit Fluorometer) and not your stock solution.
9.If you are comparing Qubit assay results to concentration obtained by UV absorbance, and the concentration based on absorbance is significantly higher, it may be because of nucleic acid or protein contamination. The Qubit assays are much more specific for DNA, RNA, or protein than absorbance readings.
The value is decreasing over time when using the Qubit Fluorometer. What could be causing this?
Please see our suggestions below:
  • Make sure that you take your reading only after incubating for at least 2 minutes (15 minutes for protein).
  • If you leave the assay tube in the Qubit Fluorometer and take multiple readings, the readings will go down as the tube heats up inside the instrument. If you want to take multiple readings, remove the tube from the instrument, place it in a tube rack, and allow it to equilibrate to room temperature for at least 30 seconds before rereading the tube.
  • You may read the sample up to 3 hours after mixing if it is protected from light. After this time, the reading will not be accurate.
  • Keep standards and sample tubes in the dark and protected from light in between readings.
    Find additional tips, troubleshooting help, and resources within ourNucleic Acid Quantification Support Center.
  • I'm trying to quantify some DNA labeled with a fluorophore. Will this work?
    PicoGreen dye and other fluorescence-based quantification reagents are not recommended for quantifying dye-conjugated nucleic acids. The attached dye molecules can interfere with either binding and/or fluorescence output of the quantification reagents.
    Does DNA length have an effect on the dsDNA assays?
    Strands that are roughly in the 20-mer range or shorter show a lower level of signal. For dsDNA samples that are composed of mostly short strands, the reagent may still be used, but one should use a dsDNA standard that is of comparable length as the sample.
    Find additional tips, troubleshooting help, and resources within our Nucleic Acid Quantification Support Center.
    What is the difference between the Quant-iT PicoGreen DNA, Quant-iT DNA, and Qubit DNA assays?
    The Qubit Fluorometer contains highly optimized algorithms that calculate the concentration of the sample using either the Qubit assays or the Quant-iT DNA assays. The Quant-iT PicoGreen DNA assay may be adapted to the Qubit Fluorometer using the MyQubit firmware. The performance of all of these assays is similar.
    The Quant-iT PicoGreen DNA assay is the most established assay and the most general-purpose (http://tools.thermofisher.com/content/sfs/manuals/PicoGreen-dsDNA-protocol.pdf). It requires the dilution of the standard DNA and buffer but can be adapted for use with either cuvettes, microplates, or the NanoDrop 3300.
    The Quant-iT DNA assays provide a ready-to-use buffer and pre-diluted standard DNA for analyzing a large number of samples (>20 samples) using a 96-well microplate with no further adaptation.
    The Qubit assays (https://www.thermofisher.com/us/en/home/industrial/spectroscopy-elemental-isotope-analysis/molecular-spectroscopy/fluorometers/qubit/qubit-assays/myqubit.html) are intended for low throughput (<20 samples), and are only used on the Qubit Fluorometer.
    Find additional tips, troubleshooting help, and resources within our Nucleic Acid Quantification Support Center.
    What are the excitation/emission wavelengths for dyes in the Qubit Assays?
    The exact excitation/emission wavelength information is proprietary. Here are the approximate excitation/emission wavelengths:
    - Qubit dsDNA HS Assay: ~500 nm/ ~530 nm
    - Qubit dsDNA BR Assay: ~510 nm/ ~530 nm
    - Qubit ssDNA Assay: ~490 nm/ ~520 nm
    - Qubit RNA HS Assay: ~640 nm/ ~670 nm
    - Qubit RNA BR Assay: ~640 nm/ ~670 nm
    - Qubit microRNA Assay: ~500 nm/ ~520 nm
    - Qubit Protein Assay: ~470 nm/ ~570 nm
    Find additional tips, troubleshooting help, and resources within our Nucleic Acid Quantification Support Center.
    Can I make my own assay for the Qubit Fluorometer?
    Yes, you can, for Qubit instruments developed after the original Qubit (1.0) Fluorometer. See MyQubit assay instructions here (http://www.thermofisher.com/us/en/home/life-science/laboratory-instruments/fluorometers/qubit/qubit-assays/myqubit.html.html).
    I have a crude lysate. Will the Quant-iT and Qubit assays work?
    Generally, the cleaner the sample the better. Some salts, proteins, and detergents are tolerated in the assays; see the specific assay protocol for which ones and at what concentrations.
    I have your Qubit ssDNA Assay Kit, but my sample has both ssDNA and dsDNA. How specific is the ssDNA kit?
    Unfortunately, the ssDNA reagent will label all types of nucleic acids. The only option would be to purify your sample prior to use with the Quibit kit for quantitation.
    How does the accuracy and sensitivity of the Qubit quantitation assays using the Qubit fluorometer compare to a microplate reader?
    The accuracy and sensitivity of the Qubit quantitation assays are the same as that of a microplate reader. This was a requirement during product development. The detection limits for each Qubit kit can be found on the corresponding product manual, which can be found by searching our website by keyword or catalog number.
    Find additional tips, troubleshooting help, and resources within our Nucleic Acid Quantification Support Center.
    Can the Qubit kits give an indication of sample quality in nucleic acid samples?
    No. The Qubit DNA and RNA kits only quantify the amount of either DNA or RNA in the sample. The Qubit fluorometer cannot take absorbance readings to provide a A260/A280 ratio or detect protein in nucleic acid samples. This can be done with the NanoDrop instrument. If your sample contains protein or other contaminants that can affect the assay, it should be further purified.
    If your sample may contain both DNA and RNA, one may use either (or both) the DNA and RNA Qubit kits and compare with samples treated with either RNase or DNase to get an accurate determination of DNA or RNA, respectively.
    Can I use the Quant-iT and Qubit Kits with other fluorometers?
    All Quant-iT and Qubit kits are compatible with all fluorometers and microplate readers that have the appropriate light sources and filters. You won't have access to the algorithm in the Qubit fluorometer for generating the standard curve provided by the instrument, instead, you must make a few dilutions of the highest standard DNA or RNA (Standard #2) in the Qubit kits to generate a standard curve with multiple data points.
    Can I use the original Quant-iT Kits with the Qubit Fluorometer?
    No, we do not recommend this. Some of the dyes in the original Quant-iT kits (those NOT listed as “for use with the Qubit fluorometer”) are not compatible with the Qubit Fluorometer. In addition, the new Quant-iT kits (labeled as “for use with the Qubit Fluorometer”) have standards formulated to be compatible with the Qubit fluorometer internal algorithms for the respective assays. The Qubit Fluorometer-compatible kits are also less expensive per assay if you are processing fewer than 20 samples at a time.
    How can I quantify the ssDNA in samples that contain both ssDNA and dsDNA, using the Qubit ssDNA Assay Kit (Cat. No. Q10212)?
    Unfortunately, it is not possible to efficiently measure the ssDNA content in a sample that also contains dsDNA with the Qubit ssDNA Assay Kit (Cat. No. Q10212). The dye in the Qubit ssDNA Assay kit binds to both ssDNA and dsDNA.
    It is possible to treat the sample with a DNase that only degrades dsDNA, such as theezDNase Enzyme (Cat. No. 11766051), and measure the ssDNA concentration afterward. However, if all dsDNA is not successfully digested, there is a risk of overestimating the ssDNA content.
    The following article describes a PCR-based method to detect ssDNA in dsDNA samples:
    Quantitative amplification of single-stranded DNA (QAOS) demonstrates that cdc13-1 mutants generate ssDNA in a telomere to centromere direction
    Find additional tips, troubleshooting help, and resources within our Nucleic Acid Purification and Analysis Support Center.
    My Qubit 4 Fluorometer, with Wifi (Cat. No. Q33238) has not installed the files to allow me to use the Qubit ssDNA Assay Kit. Could you provide the files I need?
    To access the Qubit ssDNA Assay Kit on the Qubit 4, please try selecting the Oligo option from the home screen, and the ssDNA Assay should appear there. If not, please check if you need to update your instrument firmware to the latest version v2.19, which is available for download from the Technical Resources for Qubit Fluorometers page on thermofisher.com.
    For standard/preloaded Qubit assays we do not provide individual .qbt files, but they are included in the firmware itself.