QuantiGene™ Sample Processing Kit, blood samples, 2 Plates - FAQs
QuantiGene™ Sample Processing Kit, blood samples, 2 Plates - FAQs
View additional product information for QuantiGene™ Sample Processing Kit, blood samples - FAQs (QS0110, QS0111, QS0112)
5 product FAQs found
Can samples in TRIzol solution be run in the QuantiGene Plex assay?
No, samples in TRIzol solution cannot be run in the QuantiGene Plex assay. However, RNA purified from these samples can be run in the QuantiGene Plex assay.
Can I use the QuantiGene Plex assay with bacteria?
Although the kits were not originally validated for bacteria, we have guidelines for preparing bacterial samples for QuantiGene Plex assays: http://assets.thermofisher.com/TFS-Assets/BID/Reference-Materials/preparation-bacterial-homogenates-quantigene-plex-assays-protocol.pdf
Do you offer QuantiGene Plex assay custom services where I can send my samples for you to process?
Yes, please reach out to your Technical Sales Specialist for more information or fill out this form: https://www.thermofisher.com/us/en/home/global/forms/life-science/request-quote-quantigene-assays.html
Can I detect genomic DNA using the QuantiGene Plex assay?
Under the hybridization conditions that are used for the QuantiGene Plex assay, genomic DNA remains double-stranded. Therefore, it is not possible for the probes to bind to DNA. The signal from the probe sets comes only from RNA.
Can I use other lysis buffers with QuantiGene Plex assay samples?
We only recommend the use of QuantiGene Lysis Mixture for preparing cell and blood lysates. The Lysis Mixture has been optimized to provide stability of RNA in the lysates. In addition, QuantiGene Lysis Mixture provides the appropriate buffer conditions for the overnight hybridization.