QuantiGene™ Sample Processing Kit, blood samples
Invitrogen™
QuantiGene™ Sample Processing Kit, blood samples
This QuantiGene Sample Processing Kit for blood samples contains reagents and instructions for the preparation of cell lysates from whole blood samples for use in QuantiGene Singleplex and QuantiGene Plex assays for RNA or DNA targets.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberQuantity
QS01102 Plates
QS011110 Plates
QS01125 x 10 Plates
Catalog number QS0110
Price (USD)
367.00
Each
Quantity:
2 Plates
Request bulk or custom format
Price (USD)
367.00
Each
This QuantiGene Sample Processing Kit for blood samples contains reagents and instructions for the preparation of cell lysates from whole blood samples for use in QuantiGene Singleplex and QuantiGene Plex assays for RNA or DNA targets.
  • Easy to follow instructions
  • Preserves the integrity of the RNA/DNA without degradation
  • Choose the method that matches your whole blood or dried blood spot sample
What you get
The kit contains our proprietary lysis mixture, proteinase K, and detailed instructions for preparing whole blood from heparin, citrate, or EDTA tubes, PAXgene blood RNA tubes, Tempus™ blood RNA tubes, or from dried blood spots prepared from standard anticoagulant venous blood or finger sticks.
For Research Use Only. Not for use in diagnostic procedures.
Specifications
Product LineQuantiGene
Product TypeSample Processing Kit
Quantity2 Plates
Shipping ConditionBlue Ice
FormatKit
Sample TypeBlood
Unit SizeEach
Contents & Storage
• 10 mL Lysis Mixture, store at 15°C to 30°C
• 625 μL Proteinase K (50 μg/μL), store at -20°C
Frequently asked questions (FAQs)
Can samples in TRIzol solution be run in the QuantiGene Plex assay?
No, samples in TRIzol solution cannot be run in the QuantiGene Plex assay. However, RNA purified from these samples can be run in the QuantiGene Plex assay.
Can I use the QuantiGene Plex assay with bacteria?
Although the kits were not originally validated for bacteria, we have guidelines for preparing bacterial samples for QuantiGene Plex assays: http://assets.thermofisher.com/TFS-Assets/BID/Reference-Materials/preparation-bacterial-homogenates-quantigene-plex-assays-protocol.pdf
Do you offer QuantiGene Plex assay custom services where I can send my samples for you to process?
Yes, please reach out to your Technical Sales Specialist for more information or fill out this form: https://www.thermofisher.com/us/en/home/global/forms/life-science/request-quote-quantigene-assays.html
Can I detect genomic DNA using the QuantiGene Plex assay?
Under the hybridization conditions that are used for the QuantiGene Plex assay, genomic DNA remains double-stranded. Therefore, it is not possible for the probes to bind to DNA. The signal from the probe sets comes only from RNA.
Can I use other lysis buffers with QuantiGene Plex assay samples?
We only recommend the use of QuantiGene Lysis Mixture for preparing cell and blood lysates. The Lysis Mixture has been optimized to provide stability of RNA in the lysates. In addition, QuantiGene Lysis Mixture provides the appropriate buffer conditions for the overnight hybridization.