Defined Trypsin Inhibitor
Gibco™

Defined Trypsin Inhibitor

A sterile, phosphate-buffered saline solution (1X) containing purified soybean trypsin inhibitor. Animal origin-free.Read more
Have Questions?
Catalog NumberQuantity
R007100100 mL
Catalog number R007100
Price (USD)
58.96
Special offer
Online exclusive
Ends: 27-Jun-2026
67.00
Save 8.04 (12%)
Each
Quantity:
100 mL
Price (USD)
58.96
Special offer
Online exclusive
Ends: 27-Jun-2026
67.00
Save 8.04 (12%)
Each
A sterile, phosphate-buffered saline solution (1X) containing purified soybean trypsin inhibitor. Animal origin-free.
For Research Use Only. Not for use in diagnostic procedures.
Specifications
Cell TypeMammalian Cells
Concentration1 X
Product LineCascade Biologics
Quantity100 mL
Shipping ConditionRoom Temperature
SourceAnimal origin-free
ClassificationAnimal Origin-free
Product TypeTrypsin inhibitor
SterilitySterile-filtered
pH7.4
Unit SizeEach
Contents & Storage
minus 20 degrees C

Frequently asked questions (FAQs)

Does Diploid Growth Serum-Reduced Medium (SRM) contain L-glutamine?

The basal Diploid Growth Serum-Reduced Medium already contains 6 mM L-glutamine.

Find additional tips, troubleshooting help, and resources within our Cell Culture Support Center.

Do I have to adjust my infection parameters when using Diploid Production Serum-Free Medium (SFM)?

We recommend evaluating performance with Diploid Production Serum-Free Medium using current conditions, however, multiplicity of infection, time of infection, and time of harvest may be different than with conventional media. Closely monitor cells for cytopathic effect and evaluate viral titers at multiple time points.

Find additional tips, troubleshooting help, and resources within our Cell Culture Support Center.

Can Diploid Growth Serum-Reduced Medium (SRM) be supplemented with different types of serum?

Diploid Growth Serum-Reduced Medium can be supplemented with Fetal Bovine Serum, Newborn Calf Serum, Donor Bovine Serum with Iron, or Bovine Serum.

Find additional tips, troubleshooting help, and resources within our Cell Culture Support Center.

Which cell lines can I culture in Diploid Growth Serum-Reduced Medium (SRM)?

Diploid Growth Serum-Reduced Medium has been evaluated with fibroblast cell lines, including MRC-5, WI-38, IMR-90, BS-2, and CEF cells.

Find additional tips, troubleshooting help, and resources within our Cell Culture Support Center.

How long can I use Diploid Growth Serum-Reduced Medium (SRM) supplemented with Diploid Growth Supplement?

Once supplemented, the complete Diploid Growth Serum-Reduced Medium must be used within 4 weeks.

Find additional tips, troubleshooting help, and resources within our Cell Culture Support Center.