TheraPure™ GMP Nucleotides
Thermo Scientific™

TheraPure™ GMP Nucleotides

TheraPure GMP nucleotides help accelerate mRNA vaccine and therapeutics production from preclinical development to commercialization.
Have Questions?
Modifier l'affichagebuttonViewtableView
RéférenceType de produitConcentrationQuantitéFormat
R0471SKB006UTP, solution de sodium100 mM20 mlFournitures professionnelles
R00014SKB002ATP, solution Tris200 mM5 mlFournitures professionnelles
R00014SKB005ATP, solution Tris200 mM50 mlFournitures professionnelles
R00014SKB004ATP, solution Tris200 mM500 mlFournitures professionnelles
R044SKB001ATP, solution sodique100 mM3,5 mlFournitures professionnelles
R044SKB002ATP, solution sodique100 mM17 mlFournitures professionnelles
R0441SKB003ATP, solution sodique100 mM50 mlFournitures professionnelles
R0441SKB009ATP, solution sodique100 mM500 mlFournitures professionnelles
R00015SKB002CTP, solution Tris200 mM5 mlFournitures professionnelles
R00015SKB005CTP, solution Tris200 mM50 mlFournitures professionnelles
R00015SKB004CTP, solution Tris200 mM500 mlFournitures professionnelles
R045SKB001CTP, solution de sodium100 mM3,5 mlFournitures professionnelles
R045SKB002CTP, solution de sodium100 mM17 mlFournitures professionnelles
R0451SKB003CTP, solution de sodium100 mM50 mlFournitures professionnelles
R0451SKB009CTP, solution de sodium100 mM500 mlFournitures professionnelles
R00016SKB002GTP, solution Tris200 mM5 mlFournitures professionnelles
R00016SKB005GTP, solution Tris200 mM50 mlFournitures professionnelles
R00016SKB004GTP, solution Tris200 mM500 mlFournitures professionnelles
R046SKB001GTP, solution de sodium100 mM3,5 mlFournitures professionnelles
R046SKB002GTP, solution de sodium100 mM20,5 mlFournitures professionnelles
R0461SKB003GTP, solution de sodium100 mM50 mlFournitures professionnelles
R0461SKB009GTP, solution de sodium100 mM500 mlFournitures professionnelles
R00017SKB002UTP, solution Tris200 mM5 mlFournitures professionnelles
R00017SKB005UTP, solution Tris200 mM50 mlFournitures professionnelles
R00017SKB004UTP, solution Tris200 mM500 mlFournitures professionnelles
R047SKB001UTP, solution de sodium100 mM3,5 mlFournitures professionnelles
R0471SKB002UTP, solution de sodium100 mM100 mlFournitures professionnelles
R0471SKB003UTP, solution de sodium100 mM500 mlFournitures professionnelles
R00014SKB002CATP, solution Tris200 mM5 mlAcquisition immédiate
R044SKB001CATP, solution sodique100 mM3,5 mlAcquisition immédiate
R00015SKB002CCTP, solution Tris200 mM5 mlAcquisition immédiate
R045SKB001CCTP, solution sodique100 mM3,5 mlAcquisition immédiate
R00016SKB002CGTP, solution Tris200 mM5 mlAcquisition immédiate
R046SKB001CGTP, solution de sodium100 mM3,5 mlAcquisition immédiate
R00017SKB002CSolution UTP, Solution Tris200 mM5 mlAcquisition immédiate
R047SKB001CSolution UTP, Solution de sodium100 mM3,5 mlAcquisition immédiate
Référence R0471SKB006
Prix (EUR)
-
Type de produit:
UTP, solution de sodium
Concentration:
100 mM
Quantité:
20 ml
Format:
Fournitures professionnelles
Thermo Scientific TheraPure GMP nucleoside triphosphates (NTPs) are manufactured to ICH Q7 GMP principles and supported by a comprehensive documentation package. They are suitable as core building blocks for mRNA synthesis in the development and manufacturing of mRNA therapeutics. Nucleotides ATP, CTP, GTP, and UTP are available and are supplied in sodium- or Tris-titrated solutions.

Thermo Scientific TheraPure GMP* nucleoside triphosphates (NTPs) are manufactured to ICH Q7 GMP principles and supported by a comprehensive documentation package. They are suitable as core building blocks for mRNA synthesis in the development and manufacturing of mRNA therapeutics. Nucleotides ATP, CTP, GTP, and UTP are available and are supplied in sodium- or Tris-titrated solutions.

Designed for process development and manufacturing

TheraPure GMP nucleotides help accelerate mRNA vaccine and therapeutics production from preclinical development to commercialization by providing:

  • Quality—meet critical product quality needs to accelerate therapeutic development and manufacture
  • Minimal risk—reduce risks in developing a therapeutic by utilizing proven products
  • Technical partnership—partner with expert product and technical support
  • Secure and consistent supply—count on a secure and stable supply of product at a global scale

Supported by unique quality system

TheraPure GMP nucleotides are supported by a comprehensive quality system and documentation, including:

  • Manufactured to relevant ICH Q7 GMP guidelines
  • Animal-origin–free (AOF) manufacturing processes, materials, and facilities
  • Validated manufacturing processes and analytical test methods
  • Comprehensive impurity profiles
  • Verified compendial assays where applicable
  • Product stability data
  • Tested by analytical methods following ICH Q2 guidelines whenever possible
  • Drug master file (DMF)

Two available formats

These two formats are offered to accommodate different mRNA manufacturing processes:

  • 100 mM aqueous solution with Na+ titrated to pH 7.3–7.5 with NaOH
  • 200 mM aqueous solution with Tris counterion titrated to pH 7.3–7.7 with Tris base

Modified nucleotides

TheraPure GMP N1-methylpseudo-UTP is also available.

*TheraPure GMP refers to the quality level of the raw, ancillary, or starting materials to be used for further manufacturing. TheraPure GMP products are manufactured in facilities with ISO 9001–certified quality management systems that operate in accordance with relevant good manufacturing practice (GMP) principles, as outlined in ICH Q7 or equivalent guidance documents or standards.

For Research Use or Further Manufacturing. Not for diagnostic use or direct administration into humans or animals.
Spécifications
CouleurTransparent
À utiliser avec (application)IVT Transcription, RNA Synthesis, mRNA Synthesis
FormatFournitures professionnelles
Marqueur ou colorantNon marqué
Poids moléculaire484.14 g/mol
Type de produitUTP, solution de sodium
Pureté≥ 99%
Quantité20 ml
Durée de conservationUp to 36 months after manufacturing when stored at –20°C
Concentration100 mM
Gamme de produitsTheraPure™ GMP
pH7.3 to 7.5
Unit SizeEach