Rhod-2, Tripotassium Salt, cell impermeant
Invitrogen™

Rhod-2, Tripotassium Salt, cell impermeant

Cells may be physically loaded with cell-impermeant salt forms of these indicators using patch pipet, microinjection or pinocytotic cell-loading reagent.

Have Questions?
Catalog NumberQuantity
R14220
also known as R-14220
1 mg
Catalog number R14220
also known as R-14220
Price (USD)
548.00
Each
Quantity:
1 mg
Price (USD)
548.00
Each

Labeled calcium indicators are molecules that exhibit an increase in fluorescence upon binding Ca2+. They have uses in many calcium signaling investigations, including measuring Ca2+ in cells and tissues that have high levels of autofluorescence and also for detecting Ca2+ release generated by photoreceptors and photoactivatable chelators. Cells may be physically loaded with the cell-impermeant salt forms of these indicators using patch pipette, microinjection, or our Influx™ pinocytotic cell-loading reagent (Cat. No. I14402). The fluorescence signal from these cells is generally measured using fluorescence microscopy.

Calcium Indicator (Cell-Impermeant Salts) Specifications:

  • Label (Ex/Em of Ca2+–bound form): Rhod-2 (552/581 nm)
  • Fluorescence intensity increase upon binding Ca2+: >100 fold
  • Kd for Ca2+ in the absence of Mg2+, in buffer: ∼570 nM
  • Exhibit fluorescence increase upon binding Ca2+ with little shift in wavelength

Using TPEN to Control Heavy Metal Cations

In addition, BAPTA-based indicators such as these bind various heavy metal cations (e.g., Mn2+, Zn2+, Pb2+) with substantially higher affinity than Ca2+. Perturbations to calcium measurements caused by presence of these ions can be controlled using the heavy metal-selective chelator (Cat. No. TPEN).

More Choices for Fluorescent Calcium Indicators

We offer a large selection of Molecular Probes™ calcium indicators for use in various experimental scenarios. For more information, review Fluorescent Ca2+ Indicators Excited with Visible Light—Section 19.3 in the Molecular Probes™ Handbook.

For UV-excitable Ca2+ indicators, protein-based Ca2+ indicators, conjugates of Ca2+ indicators, and for fluorescence-based indicators of other metal ions (i.e., Mg2+, Zn2+) review Indicators for Ca2+, Mg2+, Zn2+ and Other Metal Ions—Chapter 19 in the Molecular Probes™ Handbook.

For Research Use Only. Not for human or animal therapeutic or diagnostic use.

For Research Use Only. Not for use in diagnostic procedures.
Specifications
Detection MethodFluorescence
Dye TypeRhod-2
Emission Wavelength581 nm
Excitation Wavelength552 nm
Quantity1 mg
Shipping ConditionRoom Temperature
Emission Wavelength Range570 to 585 nm
Excitation Wavelength Range540 to 555 nm
For Use With (Application)Calcium Indicator
For Use With (Equipment)Fluorescence Microscope
Product TypeCalcium Indicator
Unit SizeEach
Contents & Storage
Store in freezer -5°C to -30°C and protect from light.

Citations & References (37)

Citations & References
Abstract
Caffeine inhibits inositol trisphosphate-mediated liberation of intracellular calcium in Xenopus oocytes.
Authors:Parker I, Ivorra I
Journal:J Physiol
PubMed ID:1844813
'1. Voltage-clamp recording of Ca(2+)-activated chloride currents in Xenopus oocytes was used to study the effects of caffeine on the liberation of intracellular Ca2+ induced by photo-release of inositol 1,4,5-trisphosphate (InsP3) from caged InsP3. Bath application of caffeine, at concentrations between 0.1 and 10 mM, reduced or abolished the current ... More
Cyclic ADP-ribose and calcium-induced calcium release regulate neurotransmitter release at a cholinergic synapse of Aplysia.
Authors:Mothet JP, Fossier P, Meunier FM, Stinnakre J, Tauc L, Baux G
Journal:J Physiol
PubMed ID:9518701
'1. Presynaptic injection of cyclic ADP-ribose (cADPR), a modulator of the ryanodine receptor, increased the postsynaptic response evoked by a presynaptic spike at an identified cholinergic synapse in the buccal ganglion of Aplysia californica. 2. The statistical analysis of long duration postsynaptic responses evoked by square depolarizations of the voltage-clamped ... More
Calcium permeability of the neuronal nuclear envelope: evaluation using confocal volumes and intracellular perfusion.
Authors:O'Malley DM
Journal:J Neurosci
PubMed ID:7931542
'In many calcium-imaging studies, the nuclear envelope appears to maintain a gradient of free calcium between the nucleus and cytosol. This issue was examined by loading amphibian sympathetic neurons with the calcium indicator fluo 3 via whole-cell patch clamping. Confocal optical sectioning allowed acquisition of independent calibration curves for the ... More
Imaging of calcium transients in skeletal muscle fibers.
Authors:Vergara J, DiFranco M, Compagnon D, Suarez-Isla BA
Journal:Biophys J
PubMed ID:2015378
'Epifluorescence images of Ca2+ transients elicited by electrical stimulation of single skeletal muscle fibers were studied with fast imaging techniques that take advantage of the large fluorescence signals emitted at relatively long wavelengths by the dyes fluo-3 and rhod-2 in response to binding of Ca2+ ions, and of the suitable ... More
The membrane guanylyl cyclase, retinal guanylyl cyclase-1, is activated through its intracellular domain.
Authors:Laura RP, Dizhoor AM, Hurley JB
Journal:J Biol Chem
PubMed ID:8662612
'Retinal guanylyl cyclase-1 (RetGC-1) is a membrane guanylyl cyclase found in photoreceptor outer segments. It consists of an apparent extracellular domain (ECD) linked by a single transmembrane segment to an intracellular domain (ICD). Guanylyl cyclase activating protein-2 (GCAP-2) is a Ca2+-binding protein that activates RetGC-1 in a Ca2+-sensitive manner. To ... More