Image-iT™ FX Signal Enhancer ReadyProbes™ Reagent
Invitrogen™

Image-iT™ FX Signal Enhancer ReadyProbes™ Reagent

Image-iT™ FX Signal Enhancer ReadyProbes™ Reagent dramatically reduces background fluorescence resulting from non-specific staining during secondary detection using conjugates of streptavidin or secondary antibodies.
Have Questions?
Catalog NumberQuantity
R371076 x 2.5 mL kit
Catalog number R37107
Price (USD)
301.76
Special offer
Online exclusive
Ends: 31-Dec-2026
368.00
Save 66.24
Each
Quantity:
6 x 2.5 mL kit
Price (USD)
301.76
Special offer
Online exclusive
Ends: 31-Dec-2026
368.00
Save 66.24
Each

Image-iT™ FX Signal Enhancer ReadyProbes™ Reagent dramatically reduces background fluorescence resulting from non-specific staining during secondary detection using conjugates of streptavidin or secondary antibodies. This reagent is applied directly to slides or coverslips containing fixed and permeabilized cell or tissue samples prior to staining with fluorescent probes.

  • Easy protocol – apply ready-made solution using a dropper bottle
  • Compatible with the other steps in your immunostaining protocol
  • Effective with most fluorescent labels
  • Stable at room temperature – keep handy at your scope or cell culture area

Cell imaging applications
Image-iT™ FX Signal Enhancer reagent is a highly effective product for blocking background staining that results from the nonspecific interactions of a wide variety of fluorescent dyes with cell and tissue constituents. Background staining seen with fluorescent conjugates of streptavidin and antibody conjugates are largely eliminated when the Image-iT™ FX Signal Enhancer reagent is applied to fixed and permeabilized cells prior to staining.

Suggestions for use

  • Apply Image-iT™ FX Signal Enhancer reagent directly to slides or coverslips containing cell or tissue samples prior to immune-staining.
  • After the fix/perm step, simply wash and add enough drops to cover your sample, then incubate for 30 min. and proceed with your normal staining protocol.
  • Additional blocking steps may be performed subsequent to blocking with the Image-iT™ Signal Enhancer ReadyProbes™ Reagent, as needed

For Research Use Only. Not for use in diagnostic procedures.

Specifications
FormatBottle(s)
Product LineImage-iT, ReadyProbes
Quantity6 x 2.5 mL kit
Product TypeSignal Enhancer
Solution TypeSignal Enhancer
Unit SizeEach
Contents & Storage
2.5 mL dropper bottles

Store at ≤25°C.

Frequently asked questions (FAQs)

I used Image-iT FX Signal Enhancer solution to get rid of nonspecific nuclear labeling with Alexa Fluor 568 secondary antibody, but I also saw a significant reduction in my specific mitochondrial antibody labeling. Why is this and what can I do?

The Image-iT FX Signal Enhancer reduces non-specific binding of dye conjugates by blocking positively-charged areas of cells or tissues that attract negatively-charged dyes. In cells after fixation, some positively-charged structures are the nuclei and mitochondria. Thus, you would expect to see a reduction in both mitochondrial and nuclear signal. The lower signal you see afterward is the specific mitochondrial signal; the fluorescence that was lost was the non-specific labeling.

Find additional tips, troubleshooting help, and resources within our Cell Analysis Support Center.

Can I use the Image-iT FX Signal Enhancer instead of my normal blocking solution (BSA or serum)?

No. Image-iT FX Signal Enhancer is not a protein blocker, like BSA, normal serum, or other commercial antibody blockers. Use it as a separate step to block non-specific charge-based binding of dyes to cellular components.

Find additional tips, troubleshooting help, and resources within our Cell Analysis Support Center.

I am labeling fixed and permeabilized cultured cells with an Alexa Fluor secondary antibody. My secondary-only control is showing nuclear and mitochondrial labeling, even though I did a thorough protein blocking and tried a concentration range. What can be done to minimize this non-specific binding?

This is most likely charge-based binding due to interactions of the charge on the dyes with cellular components of opposite charge. This can be blocked by using the Image-iT FX Signal Enhancer Solution which eliminates non-specific binding due to charge. The Signal Enhancer is applied as a pre-blocking step and your regular blocking regimen should be used to limit non-specific binding due to protein-protein interactions. Signal Enhancer cannot be used on live cells.

Find additional tips, troubleshooting help, and resources within our Cell Analysis Support Center.

Can I use the ReadyProbes reagents for flow cytometry?

This is not recommended. The ReadyProbes reagents were developed for imaging applications whereas the Ready Flow reagents were optimized for flow cytometry.

Find additional tips, troubleshooting help, and resources within our Cell Analysis Support Center.

I am observing high background when I analyze my click-labeled samples. What is causing this and what can I do to reduce the background?

The click reaction is very selective between an azide and alkyne. No other side reactions are possible in a biological system. Any non-specific background is due to non-covalent binding of the dye to various cellular components. The Select FX Signal Enhancer is not effective at reducing non-specific charge-based binding of dyes following the click reaction; we do not recommend its use with the Click-iT detection reagents. The best method to reduce background is to increase the number of BSA washes. You should always do a no-dye or no-click reaction control under the same processing and detection conditions to verify that the background is actually due to the dye and not autofluorescence. You can also perform the complete click reaction on a carrier solvent-only, no EdU or no-EU control to verify the specificity of the click reaction signal.

Find additional tips, troubleshooting help, and resources within our Cell Viability, Proliferation, Cryopreservation, and Apoptosis Support Center.

Citations & References (10)

Citations & References
Abstract
Functional scaffold-free 3-D cardiac microtissues: a novel model for the investigation of heart cells.
Authors:Desroches BR, Zhang P, Choi BR, King ME, Maldonado AE, Li W, Rago A, Liu G, Nath N, Hartmann KM, Yang B, Koren G, Morgan JR, Mende U,
Journal:Am J Physiol Heart Circ Physiol
PubMed ID:22427522
To bridge the gap between two-dimensional cell culture and tissue, various three-dimensional (3-D) cell culture approaches have been developed for the investigation of cardiac myocytes (CMs) and cardiac fibroblasts (CFs). However, several limitations still exist. This study was designed to develop a cardiac 3-D culture model with a scaffold-free technology ... More
Generation of an induced pluripotent stem cell line (TRNDi008-A) from a Hunter syndrome patient carrying a hemizygous 208insC mutation in the IDS gene.
Authors:Hong J, Xu M, Li R, Cheng YS, Kouznetsova J, Beers J, Liu C, Zou J, Zheng W
Journal:Stem Cell Res
PubMed ID:31071499
'Mucopolysaccharidosis Type II (MPS II), also known as Hunter syndrome, is a rare X-linked genetic disease caused by mutations in the IDS gene encoding iduronate 2-sulfatase (I2S). This is a multisystem disorder with significant variation in symptoms. Here, we document a human induced pluripotent stem cell (iPSC) line generated from ... More
Three-dimensional residual channel attention networks denoise and sharpen fluorescence microscopy image volumes.
Authors:
Journal:Nat Methods
PubMed ID:34059829
The aberrant upregulation of exon 10-inclusive SREK1 through SRSF10 acts as an oncogenic driver in human hepatocellular carcinoma.
Authors:
Journal:Nat Commun
PubMed ID:35296659
HIV-infected macrophages resist efficient NK cell-mediated killing while preserving inflammatory cytokine responses.
Authors:
Journal:Cell Host Microbe
PubMed ID:33571449