Search
View additional product information for ReadyProbes™ Tissue Autofluorescence Quenching Kit - FAQs (R37630)
4 product FAQs found
The volume for one drop from the ReadyProbes dropper bottles is approximately 41 µL.
No. The ReadyProbes Tissue Autofluorescence Quenching Kit minimizes autofluorescence created by aldehyde fixation, red blood cell autofluorescence, and autofluorescence of collagen and elastin. It will not quench natural pigments or other autofluorescent entities.
For the ReadyProbes Tissue Autofluorescence Quenching Kit to work, the components need to be mixed in the recommended order. Component A needs to be mixed with Component B, followed by mixing in Component C.
Once the 3 components are mixed together, we recommend using this mixture within 1 hour. The mixture should not be stored beyond 1 hour.