Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          • Primary Antibodies ›
          • V5 Tag Antibodies

          Invitrogen

          V5 Tag Monoclonal Antibody (SV5-Pk1)

          Advanced Verification
          92 Published Figures
          1,653 References
          View all (58) V5 Tag antibodies

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}

          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Tech Support
          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Tech Support
          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Tech Support

          Cite V5 Tag Monoclonal Antibody (SV5-Pk1)

          Additional Information:
          {{banner.description}}
          {{$ctrl.isSkuNotesCollapsed ? 'View more' : 'View less'}}
          • Antibody Testing Data (4)
          • Published Figures (92)
          • Advanced Verification (1)
          V5 Tag Antibody in Immunocytochemistry (ICC/IF)
          Group 53 Created with Sketch.
          V5 Tag Antibody in Immunocytochemistry (ICC/IF)
          Group 53 Created with Sketch.

          FIGURE: 1 / 97

          {{ $ctrl.currentElement.advancedVerification.fullName }}
          {{ $ctrl.currentElement.advancedVerification.text }}

          V5 Tag Antibody (R960CUS) in ICC/IF

          Immunofluorescent analysis of V5 Tag was performed using 70% confluent V5-H3-6XHis transfected HEK-293 cells. The cells were fixed with 4% paraformaldehyde for 10 minutes, permeabilized with 0.1% Triton™ X-100 for 15 minutes, and blocked with 2% BSA for 1 hour at room temperature. The cells were labeled with V5 Tag Monoclonal Antibody (Product # R960-25) at 1:100 dilution and Histone H3 Recombinant Polyclonal Antibody (Product # 711055) at 0.5 µg/mL in 0.1% BSA, incubated at 4 degree celsius overnight and then labeled with Goat anti-Mouse ... View More {{ $ctrl.currentElement.advancedVerification.fullName }} validation info. View more
          Published figure supplied by benchsci-logo
          PMID: {{$ctrl.currentElement.benchSciPubmedId}}
          View Product

          {{$ctrl.videoDesc}}

          {{$ctrl.videoLongDesc}}

          V5 Tag Antibody in Immunocytochemistry (ICC/IF)
          V5 Tag Antibody in Immunocytochemistry (ICC/IF)
          V5 Tag Antibody in Western Blot (WB)
          V5 Tag Antibody in Western Blot (WB)
          V5 Tag Antibody in Western Blot, Immunocytochemistry (WB, ICC/IF)
          V5 Tag Antibody in Immunocytochemistry (ICC/IF)
          V5 Tag Antibody in Immunocytochemistry (ICC/IF)
          V5 Tag Antibody in Immunocytochemistry (ICC/IF)
          V5 Tag Antibody in Immunocytochemistry (ICC/IF)
          V5 Tag Antibody in Immunocytochemistry (ICC/IF)
          V5 Tag Antibody in Immunocytochemistry (ICC/IF)
          V5 Tag Antibody in Immunocytochemistry (ICC/IF)
          V5 Tag Antibody in Western Blot, Immunocytochemistry (WB, ICC/IF)
          V5 Tag Antibody in Immunocytochemistry (ICC/IF)
          V5 Tag Antibody in Immunocytochemistry (ICC/IF)
          V5 Tag Antibody in Western Blot, Immunocytochemistry (WB, ICC/IF)
          V5 Tag Antibody in Immunocytochemistry (ICC/IF)
          V5 Tag Antibody in Western Blot, Immunocytochemistry (WB, ICC/IF)
          V5 Tag Antibody in Immunocytochemistry (ICC/IF)
          V5 Tag Antibody in Western Blot, Immunocytochemistry (WB, ICC/IF)
          V5 Tag Antibody in Immunocytochemistry (ICC/IF)
          V5 Tag Antibody in Immunocytochemistry (ICC/IF)
          V5 Tag Antibody in Immunocytochemistry (ICC/IF)
          V5 Tag Antibody in Immunocytochemistry (ICC/IF)
          V5 Tag Antibody in Western Blot, Immunocytochemistry (WB, ICC/IF)
          V5 Tag Antibody in Immunocytochemistry (ICC/IF)
          V5 Tag Antibody in Immunocytochemistry (ICC/IF)
          V5 Tag Antibody in Immunocytochemistry (ICC/IF)
          V5 Tag Antibody in Immunocytochemistry (ICC/IF)
          V5 Tag Antibody in Immunocytochemistry (ICC/IF)
          V5 Tag Antibody in Immunocytochemistry (ICC/IF)
          V5 Tag Antibody in Immunocytochemistry (ICC/IF)
          V5 Tag Antibody in Immunocytochemistry (ICC/IF)
          V5 Tag Antibody in Immunocytochemistry (ICC/IF)
          V5 Tag Antibody in Immunocytochemistry (ICC/IF)
          V5 Tag Antibody in Immunocytochemistry (ICC/IF)
          V5 Tag Antibody in Immunocytochemistry (ICC/IF)
          V5 Tag Antibody in Western Blot, Immunocytochemistry (WB, ICC/IF)
          V5 Tag Antibody in Immunocytochemistry (ICC/IF)
          V5 Tag Antibody in Immunocytochemistry (ICC/IF)
          V5 Tag Antibody in Immunocytochemistry (ICC/IF)
          V5 Tag Antibody in Immunocytochemistry (ICC/IF)
          V5 Tag Antibody in Immunocytochemistry (ICC/IF)
          V5 Tag Antibody in Western Blot, Immunocytochemistry (WB, ICC/IF)
          V5 Tag Antibody in Western Blot, Immunocytochemistry (WB, ICC/IF)
          V5 Tag Antibody in Western Blot, Immunocytochemistry (WB, ICC/IF)
          V5 Tag Antibody in Immunocytochemistry (ICC/IF)
          V5 Tag Antibody in Immunocytochemistry (ICC/IF)
          V5 Tag Antibody in Immunocytochemistry (ICC/IF)
          V5 Tag Antibody in Immunocytochemistry (ICC/IF)
          V5 Tag Antibody in Immunocytochemistry (ICC/IF)
          V5 Tag Antibody in Immunocytochemistry (ICC/IF)
          V5 Tag Antibody in Immunocytochemistry (ICC/IF)
          V5 Tag Antibody in Immunocytochemistry (ICC/IF)
          V5 Tag Antibody in Western Blot, Immunocytochemistry (WB, ICC/IF)
          V5 Tag Antibody in Western Blot, Immunocytochemistry (WB, ICC/IF)
          V5 Tag Antibody in Immunocytochemistry (ICC/IF)
          V5 Tag Antibody in Immunocytochemistry (ICC/IF)
          V5 Tag Antibody in Immunohistochemistry (IHC)
          V5 Tag Antibody in Immunohistochemistry (IHC)
          V5 Tag Antibody in Immunohistochemistry (IHC)
          V5 Tag Antibody in Immunohistochemistry (IHC)
          V5 Tag Antibody in Immunohistochemistry (IHC)
          V5 Tag Antibody in Immunohistochemistry (IHC)
          V5 Tag Antibody in Immunohistochemistry (IHC)
          V5 Tag Antibody in Immunohistochemistry (IHC)
          V5 Tag Antibody in Immunohistochemistry (IHC)
          V5 Tag Antibody in Immunohistochemistry (IHC)
          V5 Tag Antibody in Western Blot (WB)
          V5 Tag Antibody in Western Blot (WB)
          V5 Tag Antibody in Western Blot (WB)
          V5 Tag Antibody in Western Blot (WB)
          V5 Tag Antibody in Western Blot (WB)
          V5 Tag Antibody in Western Blot (WB)
          V5 Tag Antibody in Western Blot (WB)
          V5 Tag Antibody in Western Blot (WB)
          V5 Tag Antibody in Western Blot (WB)
          V5 Tag Antibody in Western Blot (WB)
          V5 Tag Antibody in Western Blot (WB)
          V5 Tag Antibody in Western Blot (WB)
          V5 Tag Antibody in Western Blot, Immunoprecipitation (WB, IP)
          V5 Tag Antibody in Western Blot (WB)
          V5 Tag Antibody in Western Blot (WB)
          V5 Tag Antibody in Western Blot, Immunoprecipitation (WB, IP)
          V5 Tag Antibody in Western Blot (WB)
          V5 Tag Antibody in Western Blot (WB)
          V5 Tag Antibody in Western Blot (WB)
          V5 Tag Antibody in Western Blot (WB)
          V5 Tag Antibody in Western Blot (WB)
          V5 Tag Antibody in Western Blot, Immunoprecipitation (WB, IP)
          V5 Tag Antibody in Western Blot, Immunoprecipitation (WB, IP)
          V5 Tag Antibody in Western Blot, Immunoprecipitation (WB, IP)
          V5 Tag Antibody in Western Blot (WB)
          V5 Tag Antibody in Western Blot, Immunoprecipitation (WB, IP)
          V5 Tag Antibody in Western Blot (WB)
          V5 Tag Antibody in Western Blot (WB)
          V5 Tag Antibody

          Product Details

          R960CUS

          Applications
          Tested Dilution
          Publications

          Western Blot (WB)

          1:1,000-1:5,000
          View 936 publications 936 publications

          Immunohistochemistry (IHC)

          -
          View 98 publications 98 publications

          Immunohistochemistry (Paraffin) (IHC (P))

          -
          View 2 publications 2 publications

          Immunohistochemistry (PFA fixed) (IHC (PFA))

          -
          View 2 publications 2 publications

          Immunohistochemistry (Frozen) (IHC (F))

          -
          View 2 publications 2 publications

          Immunocytochemistry (ICC/IF)

          1:100-1:500
          View 272 publications 272 publications

          Flow Cytometry (Flow)

          -
          View 23 publications 23 publications

          ELISA (ELISA)

          Assay-dependent
          View 5 publications 5 publications

          Immunoprecipitation (IP)

          Assay-dependent
          View 214 publications 214 publications

          ChIP assay (ChIP)

          -
          View 49 publications 49 publications

          Functional Assay (Functional)

          -
          View 1 publication 1 publication

          Proximity Ligation Assay (PLA) (PLA)

          -
          View 10 publications 10 publications

          Dot blot (DB)

          -
          View 4 publications 4 publications

          Inhibition Assays (Inhibition)

          -
          View 1 publication 1 publication

          RNA Immunoprecipitation (RIP)

          -
          View 4 publications 4 publications

          Affinity Purification (AP)

          -
          View 2 publications 2 publications

          GST Pull Down (GST-PD)

          -
          View 2 publications 2 publications

          CUT&RUN (CUT&RUN)

          -
          View 4 publications 4 publications

          Miscellaneous PubMed (Misc)

          -
          View 22 publications 22 publications
          Product Specifications

          Species Reactivity

          Tag

          Published species

          Tag

          Host/Isotype

          Mouse / IgG2a

          Class

          Monoclonal

          Type

          Antibody

          Clone

          SV5-Pk1

          Immunogen

          V5 synthetic peptide: Gly-Lys-Pro-Ile-Pro-Asn-Pro-Leu-Leu-Gly-Leu-Asp-Ser-Thr-.

          Conjugate

          Unconjugated Unconjugated Unconjugated

          Form

          Liquid

          Purification

          purified

          Storage buffer

          PBS

          Contains

          0.01% sodium azide

          Storage conditions

          Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.

          RRID

          AB_2792973

          Product Specific Information

          NOTE: R960-25 was previously sold under product #46-0705. When re-ordering, please use #R960-25.

          R960CUS was previously sold under product #46-1157. When re-ordering, please use #R960CUS.



          This antibody recognizes amino acid sequence: -Gly-Lys-Pro-Ile-Pro-Asn-Pro-Leu-Leu-Gly-Leu-Asp-Ser-Thr-.



          This antibody is functionally tested against 20 ng of an E. coli expressed fusion protein containing a V5 epitope using a chemiluminescent substrate at a 1 minute exposure. This antibody has also been tested in Western blot against 25 ng of recombinant Positope™ protein. The Positope™ control protein is a 53 kDa recombinant protein that contains seven epitope tags, including His (C-term), HisG, c-myc, and V5. Low background was observed using chemiluminescent or alkaline phosphatase reagents for detection. For Western blot, dilute in PBS or Tris-Buffered Saline (TBS) containing 0.05% Tween-20 and 5% nonfat, dry milk (PBSTM or TBSTM).



          Using chemiluminescence as the detection method, no cross-reactivity has been observed in bacterial lysates. In mammalian lysates, a few cross-reactive proteins have been observed upon overexposure of blots.



          This antibody has also been used successfully to immunoprecipitate fusion proteins that contain theV5 epitope.

          Target Information

          Epitope tags provide a method to localize gene products in a variety of cell types, study the topology of proteins and protein complexes, identify associated proteins, and characterize newly identified, low abundance or poorly immunogenic proteins when protein specific antibodies are not available. The V5 epitope tag is derived from a small epitope (Pk) present on the P and V proteins of the paramyxovirus of simian virus (SV5). The V5 tag is usually used with all 14 amino acids (GKPIPNPLLGLDST), although it has also been used with a shorter 9 amino acid sequence (IPNPLLGLD).

          For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.

          Bioinformatics

          Protein Aliases: simian virus 5 antibody; SPV5gp2; sv5; v-5; V5; V5 Epitope Tag

          View more View less

          It has to be done as per old AB suggested Products section.

          Disclaimer

          Close

          Clicking the images or links will redirect you to a website hosted by BenchSci that provides third-party scientific content. Neither the content nor the BenchSci technology and processes for selection have been evaluated by us; we are providing them as-is and without warranty of any kind, including for use or application of the Thermo Fisher Scientific products presented.

          guarantee_icon

          Performance Guarantee

          If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*

          Learn more
          help_icon

          We're here to help

          Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.

          Contact tech support
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          • Social Media
          • Contact Us
          • Report a Site Issue
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Responsibility Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Chile flag icon
          Chile

          Your items have has been added!


          Host server : magellan-search-blue-67655d8755-x75fr:80/100.66.77.4:80.
          git-commit: dddaa802cd395f65bf1581942d0e97a089c38f41
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.46.0-Offline