Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          • Primary Antibodies ›
          • Cytokeratin Pan Type II Antibodies

          NeoBiotechnologies

          Cytokeratin, Type II (Epithelial Marker) Recombinant Rabbit Monoclonal Antibody (KRTH/4392R)

          View all (37) Cytokeratin Pan Type II antibodies

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}

          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Tech Support
          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Tech Support
          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Tech Support

          Cite Cytokeratin, Type II (Epithelial Marker) Recombinant Rabbit Monoclonal Antibody (KRTH/4392R)

          Additional Information:

          {{$ctrl.isSkuNotesCollapsed ? 'View more' : 'View less'}}
          • Antibody Testing Data (1)
          Cytokeratin, Type II (Epithelial Marker) Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          Group 53 Created with Sketch.
          Cytokeratin, Type II (Epithelial Marker) Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          Group 53 Created with Sketch.

          FIGURE: 1 / 1

          {{ $ctrl.currentElement.advancedVerification.fullName }}
          {{ $ctrl.currentElement.advancedVerification.text }}

          Cytokeratin, Type II (Epithelial Marker) Antibody (RBM53-7293-P1ABX) in IHC (P)

          Formalin-fixed, paraffin-embedded human skin stained with CK Type II Recombinant Rabbit Monoclonal Antibody (KRTH/4392R). HIER: Tris/EDTA, pH9.0, 45 min. 2°C: HRP-polymer, 30 min. DAB, 5 min. {{ $ctrl.currentElement.advancedVerification.fullName }} validation info. View more
          Published figure supplied by benchsci-logo
          PMID: {{$ctrl.currentElement.benchSciPubmedId}}
          View Product

          {{$ctrl.videoDesc}}

          {{$ctrl.videoLongDesc}}

          Cytokeratin, Type II (Epithelial Marker) Antibody in Immunohistochemistry (Paraffin) (IHC (P))

          Please note: We are reviewing Western blot images included in the antibody testing data in our catalog, including those provided by third parties. Unless expressly labeled or annotated as “raw-unedited”, Western blot images included in the antibody testing data in our catalog may have been edited, optimized or otherwise adjusted for presentation.

          Cytokeratin, Type II (Epithelial Marker) Recombinant Rabbit Monoclonal Antibody (KRTH/4392R)

          Product Details

          RBM53-7293-P1ABX

          Applications
          Tested Dilution
          Publications

          Immunohistochemistry (Paraffin) (IHC (P))

          1-2 µg/mL
          -
          Product Specifications

          Species Reactivity

          Human

          Host/Isotype

          Rabbit / IgG, kappa

          Expression System

          HEK293 cells

          Class

          Recombinant Monoclonal

          Type

          Antibody

          Clone

          KRTH/4392R

          Immunogen

          Synthetic peptide corresponding to Type II Cytokeratins residues within aa300-400 of Type II Cytokeratins
          3D Epitope / Immunogen

          Conjugate

          Unconjugated Unconjugated Unconjugated

          Form

          Liquid

          Concentration

          1 mg/mL

          Amount

          100 µg

          Purification

          Protein A/G

          Storage buffer

          PBS

          Contains

          no preservative

          Storage conditions

          -20°C or -80°C if preferred

          Shipping conditions

          Ambient (domestic); Wet ice (international)

          Product Specific Information

          Positive Control:skin or squamous cell carcinoma.

          Cellular Location: Cytoplasm.

          Target Information

          Cytokeratin pan is part of a subfamily of intermediate filament proteins that are characterized by remarkable biochemical diversity, and represented in human epithelial tissues by at least 20 different polypeptides. Cytokeratins range in molecular weight between 40 kDa- 68 kDa, and an isoelectric pH between 4.9-7.8. The individual human cytokeratins are numbered 1 to 20. The various epithelia in the human body usually express cytokeratins which are not only characteristic of the type of epithelium, but also related to the degree of maturation or differentiation within an epithelium. Cytokeratin subtype expression patterns are used to an increasing extent in the distinction of different types of epithelial malignancies. The cytokeratin antibodies are not only of assistance in the differential diagnosis of tumors using immunohistochemistry on tissue sections, but are also a useful tool in cytopathology and flow cytometric assays. The composition of cytokeratin pairs vary with the epithelial cell type, stage of differentiation, cellular growth environment, and disease state. Many studies have shown the usefulness of keratins as markers in cancer research and tumor diagnosis.

          For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.

          References (0)

          Have you cited this product in a publication?
          Let us know so we can reference it here.
          Cite this product

          Bioinformatics

          Protein Aliases: 58 kda cytokeratin; 65 kDa cytokeratin; 67 kDa cytokeratin; CK-1; CK-2P; CK-3; CK-5; CK-7; cytokeratin 1; cytokeratin 2; cytokeratin 3; cytokeratin 4; cytokeratin 6A; cytokeratin 6C; cytokeratin 6D; cytokeratin 7; cytokeratin 8; cytokeratin-2e; cytokeratin-2P; cytokeratin-5; cytokeratin-72; cytokeratin-8; cytoskeletal protein; epidermolysis bullosa simplex 2 Dowling-Meara/Kobner/Weber-Cockayne types; epidermolytic hyperkeratosis 1; epithelial keratin-2e; hair alpha protein; intermediate filament protein; K7; K2P; K76; keratin 1, type II; keratin 2, type II; keratin 2p; keratin 3, type II; keratin 4, type II; keratin 5 (epidermolysis bullosa simplex, Dowling-Meara/Kobner/Weber-Cockayne types); keratin 5, type II; keratin 6, inner root sheath, 2; keratin 6A, , type II; keratin 6A, type II; keratin 7, type II; keratin 72, type II; keratin 76, type II; keratin 8, type II; keratin K5; keratin protein K6irs; keratin type II; keratin, 55K type II cytoskeletal; keratin, epidermal type II, K6A; keratin, simple epithelial type I, K7; keratin-2 epidermis; keratin-2e; KRT5; MGC116967; MGC116968; pan keratin; sarcolectin; type II inner root sheath-specific keratin-K6irs2; type II intermediate filament protein; type II keratin subunit protein; type II mesothelial keratin K7; type-II keratin Kb1; type-II keratin Kb2; type-II keratin Kb3; type-II keratin Kb35; type-II keratin Kb4; type-II keratin Kb5; type-II keratin Kb6; type-II keratin Kb7; type-II keratin Kb8; type-II keratin Kb9; unnamed protein product

          View more View less

          Gene Aliases: AEI2; CARD2; CK-2e; CK-4; CK-6C; CK-6E; CK-72; CK-8; CK1; CK3; CK4; CK5; CK6A; CK6C; CK6D; CK7; CK8; CYK4; CYK8; DDD; DDD1; EBS1; EBS2; EBS2A; EBS2B; EBS2C; EBS2D; EBS2E; EBS2F; EHK; EHK1; EPPK; HUMCYT2A; K1; K2C7; K2C8; K2e; K3; K4; K5; K6A; K6C; K6D; K6irs; K6IRS2; K7; K72; K8; KO; KRT1A; KRT2A; KRT2B; KRT2E; KRT2P; KRT5A; KRT6; KRT6C; KRT6D; KRT6IRS2; KRTE; MECD2; NEPPK; PC3; SCL; WSN1

          View more View less

          UniProt ID: (Human) P04264, (Human) P35908, (Human) P12035, (Human) P19013, (Human) P13647, (Human) P02538, (Human) P08729, (Human) Q14CN4, (Human) Q01546, (Human) P05787

          View more View less

          Entrez Gene ID: (Human) 3848, (Human) 3849, (Human) 3850, (Human) 3851, (Human) 3852, (Human) 3853, (Human) 3855, (Human) 140807, (Human) 51350, (Human) 3856

          View more View less

          Function(s)
          protein binding carbohydrate binding structural constituent of epidermis signaling receptor activity protein heterodimerization activity structural constituent of cytoskeleton cytoskeletal protein binding scaffold protein binding
          Process(es)
          cytoskeleton organization epithelial cell differentiation keratinization intermediate filament organization negative regulation of epithelial cell proliferation morphogenesis of an epithelium positive regulation of cell proliferation cell differentiation negative regulation of cell migration killing of cells of other organism wound healing defense response to Gram-positive bacterium antimicrobial humoral immune response mediated by antimicrobial peptide negative regulation of entry of bacterium into host cell sarcomere organization epithelium development negative regulation of hepatocyte apoptotic process complement activation, lectin pathway response to oxidative stress fibrinolysis regulation of angiogenesis protein heterotetramerization cornification epidermis development keratinocyte activation keratinocyte proliferation positive regulation of epidermis development keratinocyte migration intermediate filament cytoskeleton organization response to mechanical stimulus regulation of cell migration regulation of protein localization intermediate filament polymerization
          It has to be done as per old AB suggested Products section.

          Disclaimer

          Close

          Clicking the images or links will redirect you to a website hosted by BenchSci that provides third-party scientific content. Neither the content nor the BenchSci technology and processes for selection have been evaluated by us; we are providing them as-is and without warranty of any kind, including for use or application of the Thermo Fisher Scientific products presented.

          guarantee_icon

          Performance Guarantee

          If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*

          Learn more
          help_icon

          We're here to help

          Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.

          Contact tech support
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Spain flag icon
          Spain

          TEST

          Your items have has been added!


          Host server : magellan-search-blue-74d7b4b4d9-p95z8:80/100.66.74.151:80.
          git-commit: 7aafa54bd143a040505a257d2a3ebda4095a617f
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.56.0-Offline