Search
Search
Invitrogen
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}
Highest antigen sequence indentity to the following orthologs: Mouse (95%), Rat (95%).
This recombinant protein control fragment may be used for blocking experiments with the corresponding antibody, PA5-62758. In IHC/ICC and WB experiments, we recommend a 100x molar excess of the protein fragment control based on the concentration and the molecular weight. Pre-incubate the antibody-protein control fragment mixture for 30 min at room temperature.
Essential for the coupling of ERK cascade activation with the expression of ETS family genes in megakaryocytic differentiation.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: calcineurin-like EF hand protein 3; NHE-1 interacting protein; unnamed protein product
Gene Aliases: CHP3; TSC
UniProt ID: (Human) Q96BS2
Entrez Gene ID: (Human) 54997
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support