Search
Search
Invitrogen
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}
It is recommended that the protein be aliquoted for optimal storage.
It is recommended that sterile water (1785.71428571429 µL) be added to the vial to prepare a stock solution of 0.56 mg/mL.
Concentration is measured by UV.
LILRA6 may act as receptor for class I MHC antigens.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: ILT5 019; ILT5 020; ILT5 021; ILT5 025; ILT5 030; ILT5 031; ILT5 032; ILT5 033; ILT5 034; immunoglobulin superfamily member; immunoglobulin-like transcript 5; immunoglobulin-like transcript 8; leucocyte Ig-like receptor A6; leukocyte Ig-like receptor; leukocyte immunoglobulin-like receptor, subfamily A (with TM domain), member 6; leukocyte immunoglobulin-like receptor, subfamily B (with TM and ITIM domains), member 6
Gene Aliases: CD85b; ILT-8; ILT5; ILT8; LILRB6
UniProt ID: (Human) Q6PI73
Entrez Gene ID: (Human) 79168
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support