Search
Search
Invitrogen
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}
It is recommended that the protein be aliquoted for optimal storage.
Reconstitute with sterile deionized water to 0.25mg/mL
Concentration is measured by UV.
This gene is a member of the leukocyte immunoglobulin-likereceptor (LIR) family, which is found in a gene cluster atchromosomal region 19q13.4. The encoded protein belongs to thesubfamily B class of LIR receptors which contain two or fourextracellular immunoglobulin domains, a transmembrane domain, andtwo to four cytoplasmic immunoreceptor tyrosine-based inhibitorymotifs (ITIMs). Several other LIR subfamily B receptors areexpressed on immune cells where they bind to MHC class I moleculeson antigen-presenting cells and inhibit stimulation of an immuneresponse. Multiple transcript variants encoding different isoformshave been found for this gene.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: CD85 antigen-like family member C; leucocyte Ig-like receptor B5; leukocyte immunoglobulin like receptor B5 long splicing isoform; leukocyte immunoglobulin like receptor B5 short splicing isoform; leukocyte immunoglobulin-like receptor 8; leukocyte immunoglobulin-like receptor, subfamily B (with TM and ITIM domains), member 5; unnamed protein product
Gene Aliases: CD85C; LIR-8; LIR8
UniProt ID: (Human) O75023
Entrez Gene ID: (Human) 10990
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support