SBFI, AM, cell permeant
Invitrogen™

SBFI, AM, cell permeant

Sodium-sensitive molecule used to estimate Na+ gradients in isolated mitochondria, to measure intracellular NA+ levels, or to measure Na+ efflux in cells

Have Questions?
Catalog NumberQuantity
S12631 mg
Catalog number S1263
Price (USD)
854.00
Each
Quantity:
1 mg
Price (USD)
854.00
Each

SBFI is a sodium-sensitive molecule used to estimate Na+ gradients in isolated mitochondria, to measure intracellular NA+ levels, to measure Na+ efflux in cells, and, in combination with other fluorescent indicators used to correlate changes in intracellular Na+ with Ca2+ and Mg2+ concentrations, intracellular pH, and membrane potential. Although the selectivity of SBFI for Na+ is less than that of calcium indicators such as fura-2, it is sufficient for the detection of physiological concentrations of Na+ in the presence of other monovalent cations. The spectral response of SBFI upon ion binding permit excitation ratio measurements, and this indicator can be used with the same optical filters and equipment used for fura-2.

Fluorescent Ion Indicators Specifications:

  • Label (Ex/Em): SBFI (∼340,380/500 nm)
  • Lyophilized product may be dissolved in DMSO for use
  • Product is typically loaded into cells by adding the dissolved indicator to medium containing cells

    Selectivity Considerations and Cell Loading Strategies

The dissociation constant (Kd) of SBFI for Na+ is 3.8 mM in the absence of K+, and 11.3 mM in solutions with a combined Na+ and K+ concentration of 135 mM (which approximates physiological ionic strength). SBFI is ∼18-fold more selective for Na+ than for K+.

SBFI is available as both cell-impermeant acid salt (Cat. No. S1262) and as cell-permeant acetoxymethyl (AM) esters (Cat. No. S1263), (Cat. No. S1264). The anionic acid forms can be loaded into cells using our Influx™ pinocytic cell-loading reagent (Cat. No. I14402), Chelators, Calibration Buffers, Ionophores and Cell-Loading Reagents—Section 19.8), or by microinjection, patch-pipette infusion or electroporation. For AM ester loading (Loading and Calibration of Intracellular Ion Indicators—Note 19.1), addition of the Pluronic™ F-127 (Cat. No. P3000MP), (Cat. No. P6866), (Cat. No. P6867) or PowerLoad™ (Cat. No. P10020) dispersing agents as well as relatively long incubation times—up to four hours—are typically necessary.

Find Fluorescent Indicators for Na+ and K+

We offer a number of fluorescent indicators for measuring Na+ and K+. Review Fluorescent Na+ and K+ Indicators—Section 21.1 in the Molecular Probes™ Handbook for more information on these products.

For Research Use. Not for human or animal therapeutic or diagnostic use.

For Research Use Only. Not for use in diagnostic procedures.
Specifications
Detection MethodFluorescence
Dye TypeSBFI
Emission Wavelength500 nm
Excitation Wavelength340, 380 nm
Quantity1 mg
Shipping ConditionRoom Temperature
For Use With (Application)Sodium Indicator
For Use With (Equipment)Fluorescence Microscope
Product TypeSBFI AM
Unit SizeEach
Contents & Storage
Store in freezer -5°C to -30°C and protect from light.

Citations & References (281)

Citations & References
Abstract
Glutamate release evoked by glutamate receptor agonists in cultured chick retina cells: modulation by arachidonic acid.
Authors:Duarte CB,Santos PF,Sánchez-Prieto J,Carvalho AP
Journal:Journal of neuroscience research
PubMed ID:8739156
Cultured chick-embryo heart cells respond differently to ouabain as measured by the increase in their intracellular Na+ concentration.
Authors:Ahlemeyer B, Weintraut H, Schoner W
Journal:Biochim Biophys Acta
PubMed ID:1420320
Using digital imaging microscopy with the sodium-sensitive fluorescent indicator sodium-binding benzofuran isophtalate (SBFI), we examined the cytosolic free sodium ion concentration ([Na+]i) in single chick-embryo heart cells. The distribution of the [Na+]i was homogeneous within one cell, but we found a wide cell to cell variation in the range of ... More
Effect of vasopressin on Na+ kinetics in cultured rat vascular smooth muscle cells.
Authors:Okada K, Ishikawa S, Saito T
Journal:Biochem Biophys Res Commun
PubMed ID:2124111
The effect of arginine vasopressin (AVP) on Na+ kinetics was examined in cultured rat vascular smooth muscle cells (VSMC) and rat renal papillary collecting tubule cells (RPCT) by the direct measurement of intracellular sodium concentration [(Na+]i) using fluorescence dye; SBFI. AVP increased [Na+]i in a dose-dependent manner at a concentration ... More
Mechanism of collagen activation in human platelets.
Authors:Roberts DE, McNicol A, Bose R
Journal:J Biol Chem
PubMed ID:14981087
The mechanism of collagen-induced human platelet activation was examined using Ca2+, Na+, and the pH-sensitive fluorescent dyes calcium green/fura red, sodium-binding benzofuran isophthalate, and 2',7'-bis(2-carboxyethyl)-5(6)-carboxyfluorescein. Administration of a moderate dose of collagen (10 microg/ml) to human platelets resulted in an increase in [Ca2+](i) and platelet aggregation. The majority of this ... More
Expression of the voltage-gated sodium channel NaV1.5 in the macrophage late endosome regulates endosomal acidification.
Authors:Carrithers MD, Dib-Hajj S, Carrithers LM, Tokmoulina G, Pypaert M, Jonas EA, Waxman SG,
Journal:J Immunol
PubMed ID:17548620
'Voltage-gated sodium channels expressed on the plasma membrane activate rapidly in response to changes in membrane potential in cells with excitable membranes such as muscle and neurons. Macrophages also require rapid signaling mechanisms as the first line of defense against invasion by microorganisms. In this study, our goal was to ... More