TDE Series Ultra-Low Temperature Freezers - FAQs
TDE Series Ultra-Low Temperature Freezers - FAQs
View additional product information for TDE Series Ultra-Low Temperature Freezers - FAQs (TDE30086FD, TDE30086LD, TDE30086FA, TDE30086LA, TDE30086FV, TDE30086LV)