Vybrant™ Apoptosis Assay Kits for apoptotic and necrotic cell staining, 200 assays - FAQs
Vybrant™ Apoptosis Assay Kits for apoptotic and necrotic cell staining, 200 assays - FAQs
View additional product information for Vybrant™ Apoptosis Assay Kits for apoptotic and necrotic cell staining - FAQs (V13244, V13243, V23200)
2 product FAQs found
What are the fluorescence excitation/emission maxima for the dyes in the Membrane Permeability/Dead Cell Apoptosis Kit with YO-PRO-1 and PI for Flow Cytometry aka Vybrant Apoptosis Assay Kit #4, YO-PRO-1/Propidium Iodide)?
YO-PRO-1: 491/509 nm, bound to DNA
Propidium iodide: 535/617 nm, bound to DNA
What is the relative time-frame of apoptosis progression, and what products can I use to detect the different apoptotic events?
See Table 1 in this article (http://www.thermofisher.com/content/dam/LifeTech/migration/en/filelibrary/support/bioprobes/bioprobes-65.par.61751.file.dat/bioprobes-65-cellevent.pdf) where a Jurkat model system was induced with 10 µM camptothecin for time periods of 0 to 4 hours. It is important to note that these results were studied using a single cell type and induction system; results may differ for other experimental systems. Increased membrane permeability in apoptotic cells can be discriminated from dead cells using YO-PRO-1 dye in combination with propidium iodide or the SYTOX dead cell indicators. Other mid-apoptotic events are increased ROS production (detected with CellROX reagents, H2DCFDA), changes in cellular pH (BCECF, SNARF-1) and calcium release (Fluo-4, Fura-2, Indo-1). No single parameter defines apoptosis under any condition, so it is best to employ a multi-parametric approach when studying apoptosis.